Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF3027

Sigma-Aldrich

Anti-RdRp R2 of SARS-CoV Antibody, clone 3H2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies, primary antibodies

clone

3H2, monoclonal

mol wt

calculated mol wt 790.25 kDa
observed mol wt ~100 kDa

purified by

using protein G

species reactivity

SARS coronavirus

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

vaccinia virus ... NS1(956533)

General description

Replicase polyprotein 1ab (UniProt: P0C6X7; also known as pp1ab, ORF1ab polyprotein) is encoded by the rep (also known as 1a-1b) gene (Gene ID: 43740578) in SARS-CoV. It is a multifunctional protein involved in the transcription and replication of viral RNAs. It also contains the proteinases responsible for the cleavages of the polyprotein. Following its synthesis, replicase polyprotein is cleaved into 15 different chains that includes several non-structural proteins, 3C-like proteinase, proofreading exoribonuclease, helicase, uridylate-specific endoribonuclease, 2′-O-methyltransferase, and an RNA-directed RNA polymerase (RdRp) (EC:2.7.7.48), which is localized in amino acids 4370-5301. Most RNA viruses, with the exception of retroviruses, require an RdRp for replication and transcription of the viral genome, hence is important for their survival. The active site of the RdRp is the most conserved and accessible region and targeting this region for inhibition of viral replication is considered as an effective therapeutic approach. RdRp catalyzes RNA-template dependent formation of phosphodiester bonds between ribonucleotides in the presence of divalent metal ions. The initiation of synthesis occurs at the 3 -end of the template in a primer-dependent or independent manner and proceeds in the 5 → 3 direction. The catalytic site of Sars-CoV-2 RdRp is composed of a catalytic subunit known as nsp12 and two accessory subunits, nsp8 and nsp7. Its structure is highly similar to the RdRp of SARS-CoV. The core RdRp domain consists of the thumb, palm, and the fingers subdomains that are primarily involved in template binding, polymerization, nucleoside triphosphate (NTP) entry and associated functions. The palm subdomain houses most of the structurally conserved elements involved in catalysis. The thumb subdomain contains residues that are involved in packing against the template RNA and stabilize the initiating NTPs on the template. The finger subdomain holds the template RNA in place and facilitates polymerization. (Ref.: Aftab, SO., et al. (2020). J. Transl. Med. 18(1); Article 275; Hillen, HS., et al. (2020). Nature. 584 (7819); 154-156; Venkataraman, S., et al. (2018). Viruses. 10(2); 76).

Specificity

Clone 3H2 is a mouse monoclonal antibody that detects RNA dependent RNA polymerase R2 of SARS‐CoV.

Immunogen

Recombinant GST-tagged RNA dependent RNA polymerase R2 of SARS‐CoV.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant SARS-CoV-2 RdRp (NSP12) protein.

Western Blotting Analysis: A 1:250 dilution of this antibody detected recombinant RdRp R2 of SARS-CoV-2.
Anti-RdRp R2 of SARS-CoV, clone 3H2, Cat. No. MABF3027, is a mouse monoclonal antibody that detects RNA-dependent RNA polymerase of SARS-CoV and is tested for use in Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service