Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF275

Sigma-Aldrich

Anti-MRP8 Antibody/ABCC11, clone 2

clone 2, from mouse

Synonym(s):

ATP-binding cassette sub-family C member 11, Multidrug resistance-associated protein 8, MRP-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCC11(85320)

General description

MRP8/ABCC11 is an interesting protein important in a wide range of cellular processes from bile acid metabolism to ear wax secretion. MRP8/ABCC11 is involved in the ATP-dependent uptake of lipophilic anions, steroid sulfates and various bile acids and plays roles in cAMP and cGMP metabolism as well. MRP8/ABCC11 is a member of the ABC transporter superfamily of proteins. MRP8/ABCC11 is a membrane bound protein and is widely expressed except in kidney, spleen and colon. MRP8/ABCC11 is also expressed in many cancers including breast cancer where it serves as a conformational marker. Interestingly, earwax is a Mendelian trait and can be wet or dry. The Arg-184 allele of MRP8/ABCC11 gene is responsible for dry ear wax (cerumen).

Immunogen

Recombinant protein corresponding to human MRP8/ABCC11.

Application

This Anti-MRP8 Antibody/ABCC11, clone 2 is validated for use in Western Blotting for the detection of MRP8/ABCC11.

Quality

Evaluated by Western Blotting in SK-BR3 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected MRP8/ABCC11 in 10 µg of SK-BR3 cell lysate.

Target description

~170 kDa observed. The calculated molecular weight of this protein is 154 kDa, it has an isoform at 150 kDa and can run higher due to glycosolation. Uncharacterized bands may be observed in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service