Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF271

Sigma-Aldrich

Anti-FPR1/2 Antibody, clone NFPR1

clone NFPR1, from mouse

Synonym(s):

Anti-Formyl Peptide Receptor 1/2, FMLP1/2, fMet-Leu-Phe receptor, fMLP receptor, N-formyl peptide receptor, FPR, N-formylpeptide chemoattractant receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$394.00

$394.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$394.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$394.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NFPR1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FPR1(2357)

Related Categories

General description

N-formy peptide receptor 1 (also known as N-formylpeptide chemoattractant receptor or fMet-Leu-Phe receptor) & 2 (also known as RFP, FMLP-R-I, Lipoxin A4 receptor, LXA4 receptor, FMLP-related receptor I, Formyl peptide receptor-like 1) are multi-pass cell membrane G protein-coupled receptors (GPCRs) encoded by the FPR1 (also known as FPR and FMLP) and FPR2 (also known as FPR2 and FMLP-R-II) gene in human, respectively. The FPR1 gene encodes the high-affinity, and FPR2 the low-affinity, receptor for N-formyl-methionyl peptides (FMLP). Both receptors mediate FMLP-induced activation of neutrophils.

Specificity

This antibody is epitope mapped, recognizing cytoplasmic c-terminal tail residues of FPR1 (305-GQDFRERLI-313) and of FPR2 (306- GQDFRERLI-314) (Riesselman, M., et al. (2007). J. Immunol. 179:2520-2531).

Immunogen

Epitope: FPR1 (305-GQDFRERLI-313) and FPR2 (306- GQDFRERLI-314)
His-tagged recombinant protein corresponding to the FPR1 (305-GQDFRERLI-313) and FPR2 (306- GQDFRERLI-314) of human FPR1/2.

Application

Anti-FPR1/2 Antibody, clone NFPR1 is an antibody against FPR1/2 for use in Western Blotting, Flow Cytometry, Immunocytochemistry, Immunoprecipitation.
Flow Cytometry Analysis: A representative lot detected FPR1/2 in human neutraphils (Riesselman, M., et al. (2007). J. Immunol. 179:2520-2531).
Immunocytochemistry Analysis: A representative lot detected FPR1/2 in human neutraphils (Riesselman, M., et al. (2007). J. Immunol. 179:2520-2531).
Immunoprecipitation Analysis: A representative lot immunoprecipitated FPR1/2 in neutraphils (Maaty, W.S., et al. (2013). JBC. 288(38):27042-27058).
Western Blotting Analysis: A representative lot detected FPR1/2 in neutraphils (Maaty, W.S., et al. (2013). JBC. 288(38):27042-27058).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected FPR1/2 in 10 µg of THP-1 cell lysate.

Target description

~38 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service