Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2326

Sigma-Aldrich

Anti-Adeno-associated Virus 9 Antibody, clone HL2374

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$590.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$590.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

clone

HL2374
monoclonal

purified by

using protein G

species reactivity

virus, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
dot blot: suitable

isotype

IgG3κ

capacity

neutralization

General description

Adeno-associated virus 9 capsids.

Specificity

Clone HL2374 is a mouse monoclonal antibody that detects adenoassociated virus 9.

Application

ELISA Analysis: A representative lot detected Adeno-associated Virus 9 in ELISA applications (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Dot Blot Analysis: A representative lot detected Adeno-associated Virus 9 in Dot Blot applications (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Neutralizing Analysis: A representative lot neutralized Adeno-associated Virus 9 infection in HeLa cells. (Tseng, Y.S., et. al. (2016). J Virol Methods. 236:105-110).

Quality

Evaluated by Dot Blot with AAV8 and AAV9 virus like particles.

Dot Blot Analysis: Various concentrations of this antibody detected AAV9, but not AAV8 like particles.

Physical form

Purified mouse monoclonal antibody IgG3 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Virus (Adeno-associated virus 9).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service