Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2109

Sigma-Aldrich

Anti-canine NKp46 (CD335) Antibody, clone 48A

Synonym(s):

CD335, Canine NKp46, NK cell-activating receptor, NK-p46, Natural cytotoxicity triggering receptor 1, Natural killer cell p46-related protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00

$231.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 μG
$231.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$231.00


In StockDetails


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

clone

48A, monoclonal

species reactivity

canine

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

Gene Information

canine ... NCR1(611390)

General description

Natural cytotoxicity triggering receptor 1 (UniProt: S4S7F8; also known as Lymphocyte antigen 94 homolog, NK cell-activating receptor, Natural killer cell p46-related protein, NK-p46, NKp46, CD335) is encoded by the NCR1 gene (Gene ID: 611390) in canine species. NK-p46 is a single-pass type I membrane protein that belongs to the natural cytotoxicity receptor (NCR) family and serves as a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis and elimination of virus-infected cells. It is considered as a primary activating receptor mediating NK cell antiviral function. It is involved in the functional recognition of viral HA and hemagglutinin-neuraminidases. Following the initiation of an NK-target cell interaction, NKp46 is reported to cluster at the cell membrane, specifically at the immune synapse where it mediates cytoskeletal rearrangement and cellular polarization. Canine NK-p46 is synthesized with a signal peptide of 20 amino acids, which is subsequently cleaved off. The mature form has an extracellular domain (aa 21-255), a helical domain (aa 256-279), and a cytoplasmic domain (aa 280-316). It also has an Ig domain (129-211). Clone 48A is shown to recognize CD3 /NKp46+ cells that display phenotypic and functional similarities to human NK cells. Also, it recognizes a subset of T cells that are NKp46 positive. (Ref.: Foltz, JA et al. (2016). Front. Immunol. 7; 521).

Specificity

Clone 48A is a mouse monoclonal antibody that specifically detects canine NKp46.

Immunogen

L-cells expressing canine NKp46 fusion protein

Application

Anti-canine NKp46 (CD335), clone 48A, Cat. No. MABF2109, is a mouse monoclonal antibody that detects canine Natural cytotoxicity triggering receptor 1(NKp46) and has been tested for use in ELISA, Flow Cytometry, and Immunocytochemistry.

ELISA Analysis: A representative lot detected canine NKp46 (CD335) in ELISA applications (Foltz, J.A., et. al. (2016). Front Immunol. 7:521).

Flow Cytometry Analysis: A representative lot detected canine NKp46 (CD335) in Flow Cytometry applications (Foltz, J.A., et. al. (2016). Front Immunol. 7:521).

Quality

Evaluated by Immunocytochemistry in canine peripheral blood mononuclear cells (PBMCs).

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected canine NKp46 (CD335) in canine peripheral blood mononuclear cells (PBMCs).

Target description

36.13 kDa calculated.

Physical form

Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service