Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2107

Sigma-Aldrich

Anti-Chlamydial LPS Antibody, clone EVI-H1

clone EVI-H1, from mouse

Synonym(s):

Chlamydial lipopolysaccharide, Chlamydia trachomatis LPS

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$695.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$695.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

EVI-H1, monoclonal

species reactivity

bacteria

packaging

antibody small pack of 25 μg

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2a

target post-translational modification

unmodified

General description

Chlamydia trachomatis is a Gram-negative bacterium that is responsible to sexually transmitted diseases leading to pelvic inflammatory disease, ectopic pregnancy, infertility, and outbreaks of trachoma-associated blindness and lymphogranuloma venereum (LGV). Chlamydia trachomatis consists of eighteen different serological variants (serovars) that include a few subvariants. These are identified based on serological reactivity of the epitopes on their outer membrane. Intracellularly chlamydia replicates within a vacuole. Chlamydia infection is initiated with the expression of a chlamydial early gene product(s), which isolate the inclusion from the endocytic-lysosomal pathway and makes it fusogenic with sphingomyelin-containing exocytic vesicles. This change in vesicular interaction allows the delivery of the vacuole to the peri-Golgi region of the host cell. Antigens from all members of the Chlamydia genus display heat resistance and sensitivity to oxidation by sodium periodate. Lipopolysaccharides (LPS) of Chlamydia consists of at least three antigen domains, two of which are shared by the LPS of certain other gram-negative organisms and one that is unique only to chlamydial LPS. It has been reported that the quantitative yields of chlamydial LPS can be achieved when elementary bodies (EBs) are first reduced and alkylated prior to extraction with hot phenol-water. (Ref.: Caldwell, HD., and Hitchcock, PJ. (1984). Infect. Immun. (44(2); 306-314; Scidmore, MA., et al. (1996). Infect. Immun. 64(12); 5366-5372; Turingan, RS et al. (2017). PLoS ONE 12(5): e0178653).

Specificity

Clone EVH-H1 detects lipopolysaccharide from Chlamydia genus.

Immunogen

Formalin-killed elementary bodies of the Chlamydia trachomatis L2 serovar.

Application

Anti-Chlamydial LPS, clone EVI-H1, Cat. No. MABF2107, is a mouse monoclonal antibody that detects Chlamydial lipopolysaccharide (LPS) and has been tested for use in Electron Microscopy, Immunocytochemistry, Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Chlamydial LPS in Immunofluorescence applications (Su, H., et. al. (2000). Infect Immun. 68(1):192-6; Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).

Immunocytochemistry Analysis: A representative lot detected Chlamydial LPS in Immunocytochemistry applications (Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).

Electron Microscopy Analysis: A representative lot detected Chlamydial LPS with Trasnsmission Electron Microscopy applications (Wolf, K., et. al. (2001). Infect Immun. 69(5):3082-91).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in HeLa cells infected with chlamydia trachomatis serovar L2 (LGV 434 L2).

Western Blotting Analysis: 2 µg/mL of this antibody detected Chlamydial LPS in HeLa cells infected with chlamydia trachomatis serovar L2 (LGV 434 L2).

Target description

~15 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service