Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF2091

Sigma-Aldrich

Anti-CD6 Antibody, clone 34-3

clone 34-3, from rat

Synonym(s):

T-cell differentiation antigen CD6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

34-3, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Cd6(12511)

Related Categories

General description

T-cell differentiation antigen CD6 (UniProt: Q61003; also known as CD6) is encoded by the Cd6 gene (Gene ID: 12511) in murine species. CD6 is a single-pass type I membrane protein that is detected at the immunological synapse the contact zone between antigen-presenting dendritic cells and T-cells. It functions as a costimulatory molecule and promotes T-cell activation and proliferation. After T-cell activation, CD6 is hyperphosphorylated on serine and threonine residues and it is also phosphorylated on tyrosine residues in response to stimulation of the TCR complex. CD6 is predominantly expressed in thymus, lymph node, and spleen. It mediates cell-cell contacts and regulates T-cell responses via its interaction with ALCAM/CD166. It also contributes to signaling cascades triggered by activation of the TCR/CD3 complex. It functions as calcium-dependent pattern receptor that binds and aggregates both Gram-positive and Gram-negative bacteria. It binds both lipopolysaccharide (LPS) from Gram-negative bacteria and lipoteichoic acid from Gram-positive bacteria. It mediates activation of the inflammatory response and the secretion of pro-inflammatory cytokines in response to LPS. CD6 is synthesized with a signal peptide (aa 1-16), which is cleaved off to produce a mature form, which has an extracellular domain (aa 17-398), a transmembrane region (aa 399-419), and a long cytoplasmic tail (aa 420-665).

Specificity

Clone 34-3 is a rat monoclonal antibody that detects T-cell differentiation antigen CD6 in murine species.

Immunogen

CD6-transfected NB2 cells
Epitope: extracellular domain

Application

Anti-CD6, clone 34-3, Cat. No. MABF2091, is a rat monoclonal antibody that detecs T-cell differentiation antigen CD6 and has been tested for use in Flow Cytometry.
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in mouse splenocytes.

Flow Cytometry Analysis: 1 µg of this antibody detected CD6 in one million mouse splenocytes.

Target description

72.26 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service