Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2081

Sigma-Aldrich

Anti-CSF-1 Antibody, clone 5A1

Synonym(s):

MCSF, Macrophage colony-stimulating factor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5A1, monoclonal

mol wt

calculated mol wt 60 kDa
observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
inhibition assay: suitable
neutralization: suitable
radioimmunoassay: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

mouse ... Csf1(12977)

Specificity

Clone 5A1 is a rat monoclonal antibody that detects Macrophage colony-stimulating factor 1 (CSF-1).

Immunogen

Purified, serum-free, stage 4 CSF-1 from murine L cell-conditioned medium.

Application

Quality Control Testing

Evaluated by ELISA with recombinant mouse CSF-1.

ELISA Analysis: Serial dilutions of this antibody detected recombinant mouse CSF-1.

Tested Applications

Neutralizing: A representative lot neutralized the colony-stimulating activity of CSF-1.(Lokeshwar, B.L., et al. (1989). J Immunol Methods. 123(1):123-9).

ELISA Analysis: A representative lot detected CSF-1 in ELISA applications (Lokeshwar, B.L., et al. (1989). J Immunol Methods. 123(1):123-9).

Radioimmunoassay: A representative lot detected CSF-1 in Radioimmunoassay applications (Lokeshwar, B.L., et al. (1989). J Immunol Methods. 123(1):123-9).

Inhibition: A representative lot bound and inhibited the biologic function of CSF-1.(Lokeshwar, B.L., et al. (1989). J Immunol Methods. 123(1):123-9).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Macrophage colony-stimulating factor 1 (UniProt: P07141; also known as CSF-1, MCSF) is encoded by the Csf1 (also known as Csfm) gene (Gene ID: 12977) in murine species. CSF-1 is a widely expressed, single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-32), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 33-492), a transmembrane domain (aa 493-515), and a cytoplasmic domain (aa 516-552). Following its synthesis, it can be cleaved into a 43 kDa subunit CSF1 and a processed secreted form. CSF-1 is a cytokine that plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells and promotes the release of pro-inflammatory chemokines, and thereby plays an important role in innate immunity and in inflammatory processes. It also plays an important role in the regulation of osteoclast proliferation and differentiation, the regulation of bone resorption, and is required for normal bone development. Its actions are mediated by a high affinity CSF1-receptor. Clone 5A1 is shown to bind and inhibit the biological functions of CSF-1. It is reported to inhibit colony formation of tissue mononuclear phagocyte colony-forming cells as well as the committed bone marrow stem cells for both granulocytes and monocytes. The extent of colony inhibition by this clone is dependent on the tissue origin of colony-forming cells. Two isoforms of CSF-1 have been described that are produced by alternative splicing. (Ref.: Wei, S., et al. (2006). J. Leukoc. Biol. 80(6); 1445-1453; Lokeshwar, BL., and Lin, H-S. (1988). J. Immunol. 41(2); 483-488).

Physical form

Purified rat monoclonal antibody IgG1 in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service