Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1786

Sigma-Aldrich

Rat IgG1 Negative Control, clone HRPN, Azide Free Antibody

clone HRPN, from rat, purified by affinity chromatography

Synonym(s):

Immunoglobulin G1, IgG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$184.00

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$184.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$184.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

clone

HRPN, monoclonal

purified by

affinity chromatography

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Application

This rat monoclonal Rat IgG1 Negative Control, clone HRPN, Azide Free, Cat. No. MABF1786 is validated for use in Flow Cytometry.

Physical form

Purified Rat monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, pH7.2. This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Other Notes

Concentration: Please refer to lot specific datasheet.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ignacio Benedicto et al.
Nature communications, 8, 15374-15374 (2017-05-20)
The outer blood-retina barrier is established through the coordinated terminal maturation of the retinal pigment epithelium (RPE), fenestrated choroid endothelial cells (ECs) and Bruch's membrane, a highly organized basement membrane that lies between both cell types. Here we study the
Yan Cheng et al.
Laboratory investigation; a journal of technical methods and pathology, 100(9), 1208-1222 (2020-05-28)
Aberrant activation of the hedgehog (Hh) signaling pathway is associated with the formation of medulloblastoma (MB), the most common malignant pediatric brain tumor. However, tumor cells from human and mouse MB can not be passaged or preserved after being adherently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service