Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1514

Sigma-Aldrich

Anti-IL-12/IL-23 p40 (mouse), PE, clone C17.8 Antibody

clone C17.8, from rat, PE

Synonym(s):

Interleukin-12 subunit beta (IL-12B), Cytotoxic lymphocyte maturation factor 40 kDa subunit (CLMF p40), IL-12 subunit p40, IL-12/IL-23 p40

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$493.00

$493.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$493.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$493.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

C17.8, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Il12B(16160)

General description

The C17.8 antibody is specific for the 40 kDa (p40) protein subunit shared by the cytokines IL-12 and IL-23. To form IL-12, p40 assembles with a separate 35 kDa protein known as p35, resulting in a 70 kDa functional cytokine. IL-12 is secreted by activated monocytes, macrophages, and dendritic cells, and has been shown to target naïve, resting CD4+ T cells to promote their proliferation and secretion of cytokines. IL-23 contains the p40 subunit in combination with a 19 kDa protein chain, p19; its primary source being activated dendritic cells and other antigen-presenting cells. IL-23 appears to target different cell types than IL-12, acting on memory CD4+ T cells to induce a strong proliferative response and contributing to the generation and expansion of Th17 cells. Like the cytokines themselves, the receptors for IL-12 and IL-23 share one subunit, as well as containing distinct cytokine-specific subunits.As the C17.8 antibody binds to a shared subunit of both IL-12 and IL-23, it may be used as a marker for either IL-12 or IL-23 expression in dendritic cells, monocytes and macrophages, and is widely used for neutralization of activity associated with either cytokine. Please choose the appropriate format for each application.
Anti-IL-12/IL-23 p40 (mouse), PE, clone C17.8 Antibody is an antibody against IL-12 for use in Flow Cytometry.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected IL-12/IL-23 p40 in mouse macrophages.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayako Hino et al.
The American journal of pathology, 164(4), 1327-1335 (2004-03-25)
IL-12 consists of two disulfide-linked subunits, p40 and p35, that form functionally active heterodimers for the induction of Th1 cells. In contrast to IL-12 heterodimers, p40 monomers and homodimers possess inhibitory effects on Th1 cells leading to the creation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service