Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF118

Sigma-Aldrich

Anti-sTRAIL Antibody, clone III6F

clone III6F, from mouse

Synonym(s):

Tumor necrosis factor ligand superfamily member 10, Apo-2 ligand, Apo-2L, TNF-related apoptosis-inducing ligand, Protein TRAIL, CD253

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

III6F, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFSF10(8743)

Related Categories

General description

TNF-related apoptosis-inducing ligand (sTRAIL or Apo-2L) is the endogenous ligand for the ubiquitous pro-apoptotic TRAIL-R1 and TRAIL-R2 receptors. Upon ligand binding, TRAIL receptors form trimeric structures at the plasma membrane around a central zinc atom. The trimeric TRAIL receptors bind the FADD adaptor protein which in turn recruits caspase 8. TRAIL/APO-2L signals ultimately converge on caspase 3 and related proteins that promote cell death. Importantly, activated TRAIL receptors work cooperatively with death receptors 4 and 5 in transformed cells to induce caspase signaling, thereby preventing the proliferation of abnormal cells. TRAIL/ApO-2L has been implicated in pathophysiology of many disease processes, including cancer and autoimmune diseases.

Immunogen

Human soluble TRAIL

Application

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected sTRAIL in frozen sections from human tonsils, lymph nodes and appendix (Liabakk, N. B., et al. (2002). J Immunol Methods. 259(1-2):119-128.).

Flow Cytometry Analysis: A representative lot from an independent laboratory detected sTRAIL in REH cells (Liabakk, N. B., et al. (2002). J Immunol Methods. 259(1-2):119-128.).
Research Category
Inflammation & Immunology
Research Sub Category
Apoptosis - Additional
This Anti-sTRAIL Antibody, clone III6F is validated for use in Flow Cytometry, IHC, Immunoprecipitation for the detection of sTRAIL.

Quality

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis: 2.5 µg/mL of this antibody detected sTRAIL in 2X10E5 HeLa cells.

Target description

33 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service