Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE978

Sigma-Aldrich

Anti-Paired box protein Pax-1 Antibody, clone 7F11.2

clone 7F11.2, from mouse

Synonym(s):

HuP48, Paired box protein Pax-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F11.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAX1(5075)

General description

Paired box protein Pax-1 (UniProt P15863; also known as HuP48) is encoded by the PAX1 (also known as HUP48, OFC2) gene (Gene ID 5075) in human. Pax-1 belongs to the family of transcription factors with conserved DNA-binding paired box domain. The family of paired box (PAX) genes are divided into subgroup I (PAX1 and PAX9), II (PAX2, PAX5, and PAX8), III (PAX3 and PAX7), and IV (PAX4 and PAX6). Pax1 & Pax9 are paralogous genes with overlapping expression and function. Pax1 is required for the proper development of the ventral structures of the vertebral column and double homozygous mutants of Pax1 and Pax9 result in a dramatic increase in the severity of the vertebral defects. During the early stages of chondrogenic differentiation, Pax1 induces and transactivates the transcriptional repressor Nk3 homeobox 2 (Nkx3.2) that promotes chondrogenesis of sclerotomal cells, while Pax1 act as a negative regulator during chondrogenic differentiation by antagonizing against Sox9-driven chondrocyte maturation. Pax1 is downregulated as cartilage formation proceeds and becomes restricted to the mesenchymal condensations that give rise to the anlagen of the intervertebral discs, and to the perichondrium surrounding the cartilaginous vertebral bodies.

Specificity

Reactivity toward spliced isoforms 2 and 3 has not been determined.

Immunogen

GST-tagged recombinant protein corresponding to the N-terminal half of human Paired box protein Pax-1.

Application

This Anti-Paired box protein Pax-1 Antibody, clone 7F11.2 is validated for use in Western Blotting for the detection of Paired box protein Pax-1.

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Paired box protein Pax-1 in 10 µg of K562 cell lysate.

Target description

~58 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service