Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE948

Sigma-Aldrich

Anti-BRE1-A (RNF20) Antibody, clone 8A4.2

clone 8A4.2, from mouse

Synonym(s):

E3 ubiquitin-protein ligase BRE1A, BRE1 E3 ubiquitin ligase homolog, BRE1-A, hBRE1, Homolog of S. cerevisiae BRE1, RING finger protein 20

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$406.00

$406.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$406.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$406.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8A4.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RNF20(56254)

General description

E3 ubiquitin-protein ligase BRE1A (UniProt Q5VTR2; also known as BRE1 E3 ubiquitin ligase homolog, BRE1-A, hBRE1, Homolog of S. cerevisiae BRE1, RING finger protein 20) is encoded by the RNF20 (also known as BRE1, BRE1A) gene (Gene ID 56254) in human. BRE1A (RNF20) is a RING finger domain-containing protein originally identified as the E3 uqibiquitin ligase that catalyzes histone H2B Lys120 monoubiquitination in yeast. The histone chaperone FACT is reported to mediate RNF20 recruitment for chromatin remodeling to initiate homologous recombination repair (HRR). RNF20 is also shown to negatively regulate hepatic fatty acid metabolism by targeting SREBP1c for ubiquitination and subsequent proteasomal degradation. In addition, RNF20 is shown to mediated the polyubiquitination of ErbB3-binding protein 1 (Ebp1) and Transcription factor activator protein 2α (AP-2α). AP-2α functions as a negative regulator of adipogenesis by suppressing the transcription of CCAAT/enhancer binding protein (C/EBPα). Overexpression of RNF20 in 3T3-L1 cells results in a down-regulation of endogenous AP-2α and a concomitant transcriptional upregulation of C/EBPα.

Immunogen

Epitope: Third coiled-coil region.
GST-tagged recombinant protein corresponding to the third coiled-coil region of human BRE1-A (RNF20).

Application

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected BRE1-A (RNF20) in 10 µg of HEK293 cell lysate.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected BRE1-A (RNF20) in 10 µg of HeLa cell lysate.

Target description

~130 kDa observed.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amila Suraweera et al.
Communications biology, 4(1), 484-484 (2021-04-21)
Genomic stability is critical for normal cellular function and its deregulation is a universal hallmark of cancer. Here we outline a previously undescribed role of COMMD4 in maintaining genomic stability, by regulation of chromatin remodelling at sites of DNA double-strand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service