Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABE870

Sigma-Aldrich

Anti-Lin28A Antibody, clone 35L33G

clone 35L33G, from mouse

Synonym(s):

Protein lin-28 homolog A, Lin-28A, Zinc finger CCHC domain-containing protein 1, Lin28A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

35L33G, monoclonal

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LIN28A(79727)

General description

Lin28 is a developmentally regulated RNA binding protein (RBP). Its down regulation is mediated in part by Lin4 and miR-125b, the mammalian micro-RNA homolog of Lin4. It is known to be negatively regulated by retinoic acid and act as a translational enhancer by driving specific mRNAs to polysomes and thus increasing the efficiency of protein synthesis by increasing the number of initiation events per mRNA molecule. Lin28 is necessary and sufficient for selectively blocking the cleavage processing of pri-let-7 miRNAs in embryonic cells (Viswanathan, 2008). Lin28 is a negative regulator of miRNA biogenesis and may play a role in blocking miRNA-mediated differentiation in stem cells and in certain cancers (Viswanathan, 2008). The mechanism by which miR-125b represses Lin28 in mammalian cells appears to involve reductions not only in translational efficiency, but also in mRNA abundance (Wu, 2005). Lin28 has shown to be essential for skeletal muscle differentiation through the translational upregulation of IGF2 expression. It is known to be expressed in testis, placenta, and embryonic stem (ES) cells.

Immunogen

Recombinant protein corresponding to human Lin28A.

Application

Anti-Lin28A Antibody, clone 35L33G is an antibody against Lin28A for use in Western Blotting, Immunoprecipitation.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Lin28A in 10 µg of mESC and NTera2 cell lysate.
Immunoprecipitation Analysis: A representative lot immunoprecipitated Lin28A in A3-1 mESC lysates (Cho, J., et al. (2012). Cell. 151:765-777).
CLIP Analysis: A representative lot immunoprecipitated Lin28A in A3-1 mESC lysates (Cho, J., et al. (2012). Cell. 151:765-777).

Quality

Evaluated by Western Blotting in H9 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Lin28A in 10 µg of H9 cell lysate.

Target description

~ 30 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service