Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABE644

Sigma-Aldrich

Anti-Cre Recombinase Antibody, clone 5B4.1

clone 5B4.1, from mouse

Synonym(s):

Recombinase cre

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5B4.1, monoclonal

species reactivity

virus

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

Escherichia coli ... Cre(2777477)

General description

Recombinase cre (UniProt P06956) is encoded by the cre gene (Gene ID 2777477) of bacteriophage P1. Cre (Causes recombination with Cyclization recombinase) is a 38 kDa DNA recombinase derived from the P1 bacteriophage. It is highly specific for a 34-bp DNA sequence motif called loxP (locus of X-over P1) found in P1 DNA. The loxP site is composed of a variable asymmetric 8-bp sequence in between two sets of 13-bp palindromic sequences (ATAACTTCGTATA-NNNTANNN-TATACGAAGTTAT). Two cre recombinase proteins, each binds to one of the two 13-bp palindromic sequences at a loxP site, form a dimer. This dimer then binds to a second dimer on another loxP site to form a tetramer. The two sites joined by the tetramer are parallel in orientation and the double stranded DNA is cut at both loxP sites. The strands are then rejoined with DNA ligase. Because the Cre gene and loxP sites are not native to most species, they are often introduced into target cells or tissues and employed as a means to precisely control the expression of genes both in cultured cells in vitro and in animals vivo.

Specificity

Clone 5B4.1 detects an epitope outside the N-terminal region.

Immunogen

GST-tagged recombinant Cre lacking the N-terminal region sequence.

Application

Anti-Cre Recombinase Antibody, clone 5B4.1, Cat. No. MABE644, is a highly specific mouse monoclonal antibody that targets Cre recombinase and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in recombinant Cre.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected 250 ng of recombinant Cre (with added N- and C-terminal tag sequences).

Target description

38.54 kDa calculated.

Linkage

Replaces: MABE3120

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service