Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE328

Sigma-Aldrich

Anti-MeCP2 Antibody, clone 4H7

clone 4H7, from rat

Synonym(s):

Methyl-CpG-binding protein 2, MeCp-2 protein, MeCp2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$564.00

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H7, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Mecp2(17257)

General description

DNA methyltransferases methylate the 5-position of cytosine in the context of CpG dinucleotides. DNA methylation is crucial for normal embryonic development, imprinting, and X chromosome inactivation. Methyl CpG binding proteins (MeCPs) specifically recognize methylated regions of DNA. It represses transcription both directly and by association with known corepressor proteins which include members of the histone deacetylase protein families.

Specificity

This antibody recognizes the C-terminus of MeCP2, which has been epitope mapped between amino acids 310-492.

Immunogen

Epitope: C-terminus
Full length, strep-tagged N-terminus of rat MeCP2.

Application

Immunoprecipitation Analysis: A representative lot was used by an an independent laboratory to detect MeCP2 in various, mouse whole brain cell lysates (Jost, K. L., et al. (2011). PLoS ONE. 6(11):e26499).
Chromatin Immunoprecipitation Analysis: A representative lot was used by an an independent laboratory to detect MeCP2 in various, mouse WT, but not in MeCP2 knockout brain nuclear extracts (Jost, K. L., et al. (2011). PLoS ONE. 6(11):e26499).
Immunocytochemistry Analysis: A representative lot was used by an an independent laboratory to detect MeCP2 in various, GFP-meCP2 transfected C2C12 cells (Jost, K. L., et al. (2011). PLoS ONE. 6(11):e26499).
Immunohistochemistry Analysis: A representative lot was used by an an independent laboratory to detect MeCP2 in various, mouse WT, but not in MeCP2 hemizygous null brain tissue sections (Jost, K. L., et al. (2011). PLoS ONE. 6(11):e26499).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This MeCP2 antibody is validated for use in WB, IHC, ICC, ChIP & IP for the detection of the MeCP2 protein

Quality

Evaluated by Western Blot in mouse brain tissue nuclear extract.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected MeCP2 in 10 µg of mouse brain tissue nuclear extract.

Target description

~75 kDa observed. Uniprot describes an isoform produced by alternative splicing at ~55 kDa The calculated molecular weight is 53 kDa, however MeCP2 has been observed between ~75-100 kDa in western blots (Jost, K. L., et al. (2011). PLoS ONE. 6(11):e26499).

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue nuclear extract

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Mahgoub et al.
Nature neuroscience, 19(11), 1506-1512 (2016-10-28)
Class I histone deacetylases (HDACs) Hdac1 and Hdac2 can associate together in protein complexes with transcriptional factors such as methyl-CpG-binding protein 2 (MeCP2). Given their high degree of sequence identity, we examined whether Hdac1 and Hdac2 were functionally redundant in
Jessica Apulei et al.
Cerebral cortex (New York, N.Y. : 1991), 29(6), 2384-2395 (2018-05-18)
The non-cell autonomous transfer of OTX2 homeoprotein transcription factor into juvenile mouse cerebral cortex regulates parvalbumin interneuron maturation and critical period timing. By analyzing gene expression in primary visual cortex of wild-type and Otx2+/GFP mice at plastic and nonplastic ages
Camille Ravel-Godreuil et al.
iScience, 24(7), 102756-102756 (2021-07-20)
Age is a major risk factor for neurodegenerative diseases like Parkinson's disease, but few studies have explored the contribution of key hallmarks of aging, namely DNA methylation changes and heterochromatin destructuration, in the neurodegenerative process. Here, we investigated the consequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service