Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE305

Sigma-Aldrich

Anti-Dnmt3b Antibody, clone D3B2-2C1

clone D3B2-2C1, from rat

Synonym(s):

DNA (cytosine-5)-methyltransferase 3B, Dnmt3b, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, M.HsaIIIB

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$186.00
100 μG
$505.00

$186.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$186.00
100 μG
$505.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$186.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D3B2-2C1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DNMT3B(1789)

General description

DNA (cytosine-5)-methyltransferase 3B (Dnmt3B) is one of three mammalian enzymes that catalyze the addition of a methyl group to the 5’ carbon of cytosine residues in CpG dinucleotides. The Dnmt3b enzyme is involved in de novo methylation and the establishment of genome-wide DNA methylation patterns during development. DNA methylation has been linked to chromosome stability and gene expression in somatic cells; and to parental imprinting and X-chromosome inactivation in germ cells. Several studies have indicated that DNA methylation patterns are altered in various cancers.

Immunogen

Linear peptide corresponding to human Dnmt3b.

Application

Anti-Dnmt3b Antibody, clone D3B2-2C1 is a Rat Monoclonal Antibody for detection of Dnmt3b also known as DNA (cytosine-5)-methyltransferase 3B & has been validated in WB & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to immunoprecipitate Dnmt3b in IP (Felle, M., et al. (2011). Nucleic Acids Res. 39(19):8355-8365.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blot in HeLa nuclear extract.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Dnmt3b in 10 µg of of HeLa nuclear extract.

Target description

~110 kDa observed. Multiple isoforms between ~80-96 kDa are known to exist. A truncated protein at ~40 kDa may be observed in some cell lysates (Ostler, K. R., et al. (2007). 26(38): 5553-5563.). Uncharacterized bands may be observed at ~20 kDa and ~34 kDa in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sadaf Harandi-Zadeh et al.
Antioxidants (Basel, Switzerland), 10(8) (2021-08-28)
Epigenetic aberrations are linked to sporadic breast cancer. Interestingly, certain dietary polyphenols with anti-cancer effects, such as pterostilbene (PTS), have been shown to regulate gene expression by altering epigenetic patterns. Our group has proposed the involvement of DNA methylation and
Katarzyna Lubecka et al.
Carcinogenesis, 37(7), 656-668 (2016-05-22)
DNA hypomethylation was previously implicated in cancer progression and metastasis. The purpose of this study was to examine whether stilbenoids, resveratrol and pterostilbene thought to exert anticancer effects, target genes with oncogenic function for de novo methylation and silencing, leading

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service