Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE2043

Sigma-Aldrich

Anti-TdT/DNTT Antibody, clone 41A

Synonym(s):

DNA nucleotidylexotransferase;EC:2.7.7.31;Terminal addition enzyme;Terminal deoxynucleotidyltransferase;Terminal transferase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

41A, monoclonal

mol wt

calculated mol wt 58.54 kDa
observed mol wt ~58 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DNTT(1791)

General description

DNA nucleotidylexotransferase (UniProt: P04053; also known as EC:2.7.7.31, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, TdT, Terminal transferase) is encoded by the DNTT (also known as TDT) gene (Gene ID: 1791) in human. DNA polymerases are essential for replication, repair, and recombination of nucleic acid. In these processes, the polymerase extends a primer using a DNA template to guide each incorporation event. However, the requirement for using a template is not universal because cells possess terminal deoxynucleotidyl transferase (TdT) that has the ability to incorporate nucleotides in a template-independent manner using only single-stranded DNA as the nucleic acid substrate. TdT catalyzes the addition of deoxynucleotides to the 3′ hydroxy terminus of either double or single stranded DNA in a template-independent manner. Although TdT preferentially adds deoxynucleotides to 3′-extensions, tailing onto 5′ overhangs or blunt ended double strands of DNA can also be achieved. TdT is present in immature pre-B and pre-T cells where it inserts N-nucleotides to the V (D) J gene segment during rearrangements of genes. It plays a vital role in the development and variation of the immune system in vertebrates. TdT is commonly used for labeling DNA fragments and vectors with homopolymer tails and/or with modified deoxynucleotides, such as biotin-dNTPs, 32P-dNTPs, or ddNTP. Three alternative splice variants of TdT have been described in human. They are designated as TdTS (short), TdTL1 (long) and TdTL2 (long). TdTL1 and TdTL2 both localize in the nucleus, however, TdTL2 is expressed more abundantly in normal small lymphocytes compared to hTdTL1, which is readily detected in transformed lymphoid cell lines. Both long isoforms possess 3 →5 exonuclease activity for nucleotide removal whereas the short isoform performs nucleotide elongation of the coding ends during V(D)J recombination. Overexpression of TdTS or TdTL2 is reported to independently reduce the efficiency of V(D)J recombination, but the simultaneous overexpression of TdTS and TdTL2 can restore normal recombination frequencies. (Ref.: Gholami, S., et al. (2017). Adv Pharm Bull. 7(2); 215-220; Motea, EA., and Berdis, AJ. (2010). Biochim. Biophys. Acta. 804(5);1151-1166).

Specificity

Clone 41A is a moue monoclonal antibody that detects human Terminal deoxynucleotidyltransferase (TdT).

Immunogen

Full-length recombinant human Terminal deoxynucleotidyltransferase (TdT).

Application

Quality Control Testing

Evaluated by Western Blotting in Human thymus tissue lysate.

Western Blotting Analysis: A 1:250 dilution of this antibody detected TdT/DNTT in Human thymus tissue extracts.

Tested Applications

Immunofluorescence Analysis: A representative lot detected TdT/DNTT in Immunofluorescence application (McClory, S., et al. (2012). J Clin Invest. 122(4); 1403-15).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected TdT/DNTT in Immunohistochemistry applications (McClory, S., et al. (2012). J Clin Invest. 122(4);1403-15).

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected TdT/DNTT in human thymus tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TdT/DNTT, clone 41A, Cat. No. MABE2043, is a mouse monoclonal antibody that detects TdT/DNTT and is tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service