Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE1923

Sigma-Aldrich

Anti-T2A Antibody, clone 5E6

Synonym(s):

Clone 5E6 Anti-T2A, T2A Detection Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$239.00
100 μG
$603.00

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$239.00
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5E6, monoclonal

purified by

using protein G

species reactivity

all

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

shipped in

2-8°C

target post-translational modification

unmodified

General description

Thosea asigna virus (TaV), a member of the genus Betatetravirus of the family Tetraviridae, is a positive-sense single-stranded RNA virus that infects lepidopteran insects. This family is divided into the Betatetravirus and Omegatetravirus genera, based upon of the appearance of the virion and genome organization. 2A peptides and 2A-like peptide sequences (known as cis-acting hydrolase elements or CHYSEL) are a superior alternative to internal ribosomal entry sites (IRES) for coordinating the expression of multiple gene products from a single recombinant construct. The 2A sequences are relatively short peptides of about 16-20 amino acids (depending on the virus of origin). They contain a highly conserved sequence, GDVEXNPGP, at the C-terminus, which is shared by different 2As and is essential for the creation of steric hindrance and ribosome skipping. These peptides allow multiple proteins to be encoded as polyproteins, which then dissociate into component proteins upon translation. The 2A sequence impairs normal peptide bond formation via a mechanism called ribosomal skipping, which results in effective, non-enzymatic generation of distinct peptide products from a single multi-cistronic construct. These peptides are used by several families of viruses, the best-known amongst these are foot-and-mouth disease virus 2A (F2A) and Thosea asigna virus 2A (T2A). Clone 5E6 is a rat monoclonal antibody that reacts with a protein in lysate from HEK293T cells transfected with various 2A-tagged proteins.

Specificity

Clone 5E6 is a rat monoclonal antibody that detects T2A tagged protein.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 17 amino acids from Thosea asigna T2A protein.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293 cells transfected with alpha-Tubulin vector.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected T2A tagged alpha-Tubulin in lysate from HEK293 cells transfected with alpha-Tubulin vector.

Tested Applications

Western Blotting Analysis: A representative lot detected T2A tagged proteins in Western Blotting applications (Lange, A., et al. (2012). Differentiation.;83(2):S105-13).

Immunofluorescence Analysis: A 1:200 dilution of this antibody detected T2A tagged proteins in mouse E14.5 lung and pancreas tissue sections (Courtesy of Dr. Ingo Burtscher, Ph.D., Helmholtz Zentrum München, Germany).

Immunohistochemistry Applications: A representative lot detected T2A tagged proteins in Immunohistochemistry applications (Lange, A., et al. (2012). Differentiation.;83(2):S105-13).

Immunofluorescence Analysis: A representative lot detected T2A tagged proteins in Immunofluorescence applications (Lange, A., et al. (2012). Differentiation.;83(2):S105-13).

ELISA Analysis: A representative lot detected T2A tagged protein in ELISA applications (Lange, A., et al. (2012). Differentiation.;83(2):S105-13).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-T2A, clone 5E6, Cat. No. MABE1923, is a rat monoclonal antibody that detects T2A and is tested for use in ELISA, Immunofluorescence, Immunohistochemistry, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service