Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1911

Sigma-Aldrich

Anti-Daxx Antibody, clone Daxx5.14

clone DAXX5.14, from mouse

Synonym(s):

Death domain-associated protein 6, hDaxx, ETS1-associated protein 1, EAP1, Fas death domain-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DAXX5.14, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DAXX(1616)

General description

Death domain-associated protein 6 (UniProt: Q9UER7; also known as Daxx, hDaxx, ETS1-associated protein 1, EAP1, Fas death domain-associated protein) is encoded by the DAXX (also known as BING2, DAP6) gene (Gene ID: 1616) in human. Daxx is a homomultimeric protein that is dispersed throughout the nucleoplasm, in PML/POD/ND10 nuclear bodies, and in nucleoli. It serves as a transcription corepressor that represses transcriptional potential of several sumoylated transcription factors. It down-regulates basal and activated transcription and its repressor activity is modulated by its recruitment to subnuclear compartments like the nucleolus or PML/POD/ND10 nuclear bodies through interactions with MCSR1 and PML, respectively. It is reported to regulate transcription in PML/POD/ND10 nuclear bodies together with PML and may thereby influence TNFRSF6-dependent apoptosis. It also acts as an adapter protein in a MDM2-DAXX-USP7 complex by regulating the RING-finger E3 ligase MDM2 ubiquitination activity. Under non-stress conditions, in association with the deubiquitinating USP7, it prevents MDM2 self-ubiquitination and enhances the intrinsic E3 ligase activity of MDM2 towards TP53 that leads to TP53 ubiquitination and proteasomal degradation. However, upon DNA damage, its association with MDM2 and USP7 is disrupted, which results in increased MDM2 autoubiquitination and consequently, MDM2 degradation and TP53 stabilization. It also acts as targeting component of the chromatin remodeling complex ATRX:DAXX that has ATP-dependent DNA translocase activity and catalyzes the replication-independent deposition of histone H3.3 in pericentric DNA repeats. However, it does not affect the ATPase activity of ATRX, but alleviates its transcription repression activity.

Specificity

Clone DAXX5.14 specifically detects human Daxx. It targets an epitope with in 40 amino acids from the C-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 318 amino acids from the C-terminal half of human Daxx.

Application

Anti-Daxx, clone Daxx5.14, Cat. No. MABE1911, is a highly specific mouse monoclonal antibody that targets Daxx and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin) and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected Daxx in Immunocytochemistry applications (Nefkens, I., et. al. (2003). J Cell Sci. 116(Pt 3):513-24; Ishov, A.M., et. al. (2002). J Virol. 76(15):7705-12).

Western Blotting Analysis: A representative lot detected Daxx in Western Blotting applications (Nefkens, I., et. al. (2003). J Cell Sci. 116(Pt 3):513-24).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunohistochemistry (Paraffin) in human testis tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Daxx in human testis tissue sections.

Target description

81.37 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonali Chaturvedi et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(29), 17240-17248 (2020-07-08)
Probabilistic bet hedging, a strategy to maximize fitness in unpredictable environments by matching phenotypic variability to environmental variability, is theorized to account for the evolution of various fate-specification decisions, including viral latency. However, the molecular mechanisms underlying bet hedging remain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service