Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1907

Sigma-Aldrich

Anti-POT1 Antibody, clone 43B7

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$239.00
100 μG
$603.00

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$239.00
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

43B7, monoclonal

mol wt

calculated mol wt 71.44 kDa
observed mol wt ~70 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... POT1(25913)

General description

Protection of telomeres protein 1 (UniProt: Q9NUX5; also known as hPot1, POT1-like telomere end-binding protein) is encoded by the POT1 gene (Gene ID: 25913) in human. POT1 is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. As a component of the shelterin complex, it is involved in the regulation of telomere length and protection. It also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintenance and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5′-TTAGGG-3′ repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5′-TAGGGTTAG-3′ sequence. Its DNA binding domains are localized to amino acids 33-48 and 270-273. POT1 contains two copies of oligonucleotide oligosaccharide-binding fold (OB-fold) domains in its N-terminal half that are responsible for single strand DNA binding activity. TPP1, a shelterin component, plays critical role in chromosome end protection and telomere length regulation and POT1 is shown to negatively regulate TPP1 phosphorylation at serine Ser255 by NIMA Related Kinase 6 (NEK6). (Ref.: Hirai, Y., et al. (2016). Genes Cells 21(8); 874-889; Hirai, Y., et al. (2012). Genes Cells. 17(3); 186-204).

Specificity

Clone 43B7 is a mouse monoclonal antibody that detects human Protection of telomeres protein 1 (POT1).

Immunogen

Full-length recombinant human Protection of telomeres protein 1 (POT1).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Protection of telomeres protein 1 (POT1) in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected POT1 in HEK293T, HepG2, and K562 cell lysates.

Immunofluorescence Analysis: A representative lot detected POT1 in Immunofluorescence application. (Hirai, Y., et al. (2012). Genes Cells. 17(3):186-204).

Immunoprecipitation Analysis: A representative lot immunoprecipitated POT1 in Immunoprecipitation application. (Hirai, Y., et al. (2012). Genes Cells. 17(3):186-204; Hirai, Y. et al. (2016). Genes Cells. (8): 874-889).
Chromatin Immunoprecipitation Analysis (ChIP): A representative lot detected POT1 in Chromatin Immunoprecipitation applications (Hirai, Y., et al. (2012). Genes Cells. 17(3):186-204).

Western Blotting using knockout lysates: A representative lot detected POT1 in wild-type, but not in KO lysates in Western Blotting application. (Hirai, Y., et al. (2012). Genes Cells. 17(3):186-204).

Western Blotting Analysis: A representative lot detected POT1 in Western Blotting application. (Hirai, Y. et al. (2016). Genes Cells. (8): 874-889).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-POT1, clone 43B7, Cat. No. MABE1907, is a mouse monoclonal antibody that detects POT1 and is tested for use in Chromatin Immunoprecipitation, Immunofluorescence, Immunoprecipitation, Western Blotting, and Western Blotting using KO lysates.

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service