Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABE1671M

Sigma-Aldrich

Anti-CCAR1 Antibody, clone 4B9.1

clone 4B9.1, from mouse

Synonym(s):

Cell division cycle and apoptosis regulator protein 1, Cell cycle and apoptosis regulatory protein 1, CARP-1, Death inducer with SAP domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B9.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CCAR1(55749)

General description

Cell division cycle and apoptosis regulator protein 1 (UniProt: Q8IX12; also known as Cell cycle and apoptosis regulatory protein 1, CARP-1, Death inducer with SAP domain, CCAR1) is encoded by the CCAR1 (also known as CARP1, DIS) gene (Gene ID: 55749) in human. CCAR1 is a cytoplasmic protein that localized to the perinuclear region. It is expressed in various epithelial cancer cell lines, including breast, colon, prostate, pancreatic and leukemia. Its expression is shown to be regulated by growth factors. Two isoforms of CCAR1 have been reported that are generated by alternative splicing. CCAR1 associates with components of the Mediator and p160 coactivator complexes that play a role as intermediaries transducing regulatory signals from upstream transcriptional activator proteins to basal transcription machinery at the core promoter. CCAR1 is recruited to endogenous nuclear receptor target genes in response to the appropriate hormone. It also functions as a p53 coactivator and may play a role in transcriptional regulation. It is reported to be involved in apoptosis signaling in the presence of the reinoid CD437. In association with CALCOCO1, it enhances GATA1- and MED1-mediated transcriptional activation from the gamma-globin promoter during erythroid differentiation of K562 erythroleukemia cells. CCAR1 can act as a coactivator and corepressor of AR-mediated transcription and contribute to chromatin looping and AR transcription complex assembly by stabilizing AR-GATA2 association on chromatin and facilitating MED1 and RNA polymerase II recruitment to AR-binding sites.

Specificity

Clone 4B9.1 detects Cell division cycle and apoptosis regulator protein 1 (CCAR1) in human cells. It targets an epitope witihin 173 amino acids from the N-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 173 amino acids from the N-terminal half of human Cell division cycle and apoptosis regulator protein 1 (CCAR1).

Application

Anti-CCAR1, clone 4B9.1, Cat. No. MABE1671, is a highly specific mouse monoclonal antibody that targets Cell division cycle and apoptosis regulator protein 1 and has been tested in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected CCAR1 in HeLa, A431, and HUVEC cells.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected CCAR1 in 10 µg of HepG2 cell lysate.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CCAR1 in 10 µg of HeLa cell lysate.

Target description

~135 kDa observed; 132.82 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service