Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE159

Sigma-Aldrich

Anti-Fox1 Antibody, clone D8F8

clone D8F8, from mouse

Synonym(s):

hexaribonucleotide binding protein 1, Ataxin-2-binding protein 1, ataxin 2-binding protein 1, Hexaribonucleotide-binding protein 1, fox-1 homolog A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

D8F8, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rbfox3(52897)

General description

Fox1, also commonly referred to as Ataxin-2-binding protein 1, is an RNA binding protein involved in the regulation of tissue-specific alternative splicing. This protein contains a highly conserved RRM (RNA recognition motif) and associates with 5′-UGCAUGU-3′ elements. This protein has been demonstrated to regulate alternative splicing of tissue specific exons and differentially spliced exons during erythropoiesis. Its alternative name, Ataxin-2-binding protein, is a result of its association with the C terminus of ataxin 2 which may contribute to the pathology of the neurogenerative disease spinocerebellar ataxia type 2 (SCA2).

Immunogen

Recombinant protein corresponding to mouse Fox1.

Application

Anti-Fox1 Antibody, clone D8F8 is a Mouse Monoclonal Antibody for detection of Fox1 also known as hexaribonucleotide binding protein 1, Ataxin-2-binding protein 1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in mouse heart tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Fox1 on 10 µg of mouse heart tissue lysate.

Target description

~55 kDa observed. Uncharacterized bands potentially due to alternative splicing have been observed at ~33 and 37 kDa

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Jacko et al.
Neuron, 97(4), 853-868 (2018-02-06)
Neuronal maturation requires dramatic morphological and functional changes, but the molecular mechanisms governing this process are not well understood. Here, we studied the role of Rbfox1, Rbfox2, and Rbfox3 proteins, a family of tissue-specific splicing regulators mutated in multiple neurodevelopmental
Yan Jiang et al.
Nature communications, 12(1), 5767-5767 (2021-10-03)
Rett syndrome (RTT) is a severe neurological disorder and a leading cause of intellectual disability in young females. RTT is mainly caused by mutations found in the X-linked gene encoding methyl-CpG binding protein 2 (MeCP2). Despite extensive studies, the molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service