Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1071

Sigma-Aldrich

Anti-BAP1 Antibody, clone 1G8

clone 1G8, from mouse

Synonym(s):

Ubiquitin carboxyl-terminal hydrolase BAP1, BRCA1-associated protein 1, Cerebral protein 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$502.00

$502.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$502.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$502.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G8, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BAP1(8314)

General description

Ubiquitin carboxyl-terminal hydrolase BAP1 (EC 3.4.19.12; UniProt Q92560; also known as BRCA1-associated protein 1, Cerebral protein 6) is encoded by the BAP1 (also known as KIAA0272, hucep-6) gene (Gene ID 8314) in human. Originally identified and named as a BRCA1-interacting protein, BAP1 is a tumor suppressor that participates in DNA-damage response, DNA synthesis and cell cycle progression. BAP1 belongs to the ubiquitin C-terminal hydrolase (UCH) family of deubiquitin enzymes (DUBs), all members of which, including UCH-L1, UCH-L3 and UCH-L5, are characterized by a conserved UCH catalytic domain (CD; BAP1 a.a. 1-235). BAP1 and UCH-L5 also share a conserved C-terminal ULD domain (a.a. 640-710 of BAP1). The region between BAP1 CD and ULD is important for binding to the HCF-1 protein. In addition, BAP1 contains a C-terminal extension (CTE; a.a. 711-729) following its ULD. Study shows that while CTE tethers BAP1 to the nucleosome core, additional interaction with ASXL1 DEUBiquitinase ADaptor domain (DEUBAD) is needed to allow BAP1 substrate processing by increasing it′s affinity towards ubiquitin. In a similar fashion, UCH-L5 activity is regulated via interaction with DEUBAD of RPN13 and INO80G. BAP1 is implicated in transcription regulation due to its interaction with transcription-related proteins, such as FOXK2, HCF-1, OGT, LSD2, MBD-6, MBD-5, ASXL1 and ASXL2. BAP1 loss leads to a variety of cancers, including metastatic cutaneous and uveal melanoma, pleural mesothelioma, renal cell carcinoma and bladder tumours.

Specificity

Clone 1G8 targets human BAP1 UCH domain.

Immunogen

Recombinant full-length human BAP1.

Application

Anti-BAP1 Antibody, clone 1G8, Cat. No. MABE1071, is a highly specific mouse monoclonal antibody that targets BAP1 and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in transfected HEK293 cells.

Western Blotting Analysis: 2 µg/mL of this antibody detected exogenously expressed human BAP1 in 10 µg lysate from transfected HEK293 cells.

Target description

~80 kDa observed. 80.36 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service