Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABD127

Sigma-Aldrich

Anti-FBXO15 Antibody, clone 3B11.1

clone 3B11.1, from mouse

Synonym(s):

F-box only protein 15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B11.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FBXO15(201456)

General description

F-box only protein 15 (UniProt Q8NCQ5) is encoded by the FBXO15 (also known as FBX15) gene (Gene ID 201456) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type, HECT-type, U-box-type, and PHD-finger-type E3 ligases. The RING finger-type E3s include seven cullin-based E3s, such as SKP1–CUL1–F-box-protein (SCF) complex and the anaphase-promoting complex/cyclosome (APC/C). F-box proteins use their N-terminal F-box motif to bind Skp1 and other motif(s) in their C-terminal region for substrates recognition and recruitment. F-box proteins are divided into the FBXW subclass that contains WD40 repeat domains, the FBXL subclass that contains leucine-rich repeat domains, and the F-box only (FBXO) subclass that contains not yet fully characterized substrate recognition domains. SCF-Fbxo15 is reported to mediate the ubiquitination of cardiolipin synthase (CLS1) for proteasomal degradation in human lung epithelial cells. PTEN-induced putative kinase 1 (PINK1) is shown to phosphorylate CLS1 and cause its release from the inner mitochondrial membrane to the outer membrane, making it accessible for Fbxo15-targeted ubiquitination.

Specificity

Clone 3B11.1 recoginzes an N-terminal epitope present in spliced isoform 1 (UniProt Q8NCQ5-1), but absent in spliced isoform 2 (UniProt Q8NCQ5-2).

Immunogen

Epitope: N-terminal region.
KLH-conjugated linear peptide corresponding an N-terminal sequence of human FBXO15.

Application

Anti-FBXO15 Antibody, clone 3B11.1 is an antibody against FBXO15 for use in Western Blotting.
Research Category
Stem Cell Research
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Quality

Evaluated by Western Blotting in human H9 embryonic stem cell lysate.

Western Blotting Analysis: A 1:250 dilution of this antibody detected FBXO15 in 10 µg of human H9 embryonic stem cell lysate.

Target description

~57 kDa observed. 57.26 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Purified mouse monoclonal IgMκ antibody in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service