Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABD117

Sigma-Aldrich

Anti-GATA Antibody3, clone 7B5

ascites fluid, clone 7B5, from mouse

Synonym(s):

Trans-acting T-cell-specific transcription factor GATA-3, GATA-binding factor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
7B5, monoclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable
western blot: suitable
citations:

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7B5, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GATA3(2625)

General description

GATA-binding factor 3 (GATA3), also known as Trans-acting T-cell-specific transcription factor GATA-3, is a transcription factor containing a zinc finger domain. GATA3 is primarily expressed in T cells, where it thought to participate in T-cell receptor gene activation through binding to enhancers. Analysis of conditional GATA3 knockout mice confirmed the critical role of GATA3 in helper T cell differentiation and proliferation. Cells expressing GATA3 develop the profile of Th2 cells, secreting IL-4, IL-5 and IL-13. Binding of GATA3 in the regulatory regions of the genes of these cytokines alters chromatin structure, increasing accessibility to other transcription factors. T cell receptor activation activates NFAT and other transcription factors that cooperate with GATA3 in inducing Th2 cell differentiation. Blocking GATA3 action has been suggested as a therapeutic strategy to treat asthma. GATA3 has also been implicated in developmental processes such as the development of the inner ear.

Immunogen

Purified recombinant fragment of human GATA3 expressed in E. Coli.

Application

Detect GATA using this mouse monoclonal antibody, Anti-GATA Antibody3, clone 7B5 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected GATA3 in human small intestine tissue.

Optimal working dilutions must be determined by end user.

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in GATA3-hIgGFc transfected HEK293 cell lysate.

Western Blot Analysis: A 1:500-2000 dilution of this antibody detected GATA3 in GATA3-hIgGFc transfected HEK293 cell lysate.

Target description

~75 kDa observed

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
GATA3-hIgGFc transfected HEK293 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service