Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABC991

Sigma-Aldrich

Anti-PD-L1 Antibody, clone 10F.9G2

clone 10F.9G2, from rat

Synonym(s):

PD-L1, PDCD1 ligand 1, Programmed death ligand 1, B7 homolog 1, B7-H1, CD274

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10F.9G2, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Related Categories

General description

Programmed cell death 1 ligand 1 (UniProt: Q9EP73; also known as PD-L1, PDCD1 ligand 1, Programmed death ligand 1, B7 homolog 1, B7-H1, CD274) is encoded by the CD274 (also known as B7h1, Pdcd1l1, Pdcd1lg1, Pdl1) gene (Gene ID: 60533) in murine species. PD-1 and PD-1 ligands 1 and 2 (PD-L1 and PD-L2) are B7:CD28 family members that regulate T cell activation and peripheral tolerance. PD-L1 is initially produced with signal peptide (aa: 1-18) sequence, the removal of which yields the mature protein with a large extracellular (aa: 19-238) region that contains an Ig-like V-type domain (aa: 19-127) and an Ig-like C2-type domain (aa: 133-225), followed by a transmembrane domain (aa: 239-259) and a cytoplasmic tail (aa: 260-290). When engaged together with the TCR, the interaction of PD-1 with its ligands delivers an inhibitory signal to T cell proliferation and cytokine production. While PD-L1 is broadly expressed in hematopoietic and non-hematopoietic cells, PD-L2 expression is highly restricted to antigen presenting cells (APCs), including dendritic cells (DCs) and macrophages. The PD-1 pathway plays a key role in the progressive loss of effector T cell responses during chronic HIV infection. Tumor-associated PD-L1 is reported to facilitate apoptosis of activated T cells and stimulate IL-10 production in T cells to mediate immune suppression. These effects can be blocked by some monoclonal PD-L1 antibodies and T cell function can be restored. (Ref.: Chen, L., and Han, X (2015). J. Clin. Invest. 125(9): 3384-3391).

Specificity

Clone 10F.9G2 specifically detects PD-L1 in murine EL4 lymphoma cells and targets the extracellular domain.

Immunogen

CHO-mPD-L1 transfectants.

Application

Detect PD-L1 using this rat monoclonal Anti-PD-L1 Antibody, clone 10F.9G2, Cat. No. MABC991. It has been tested in Flow Cytometry, Function Induction, Immunohistochemistry, and Inhibition studies.
Immunohistochemistry Analysis: A representative lot detected PD-L1 in Immunohistochemistry applications (Grabie, N., et. al. (2007). Circulation. 116(18):2062-71).

Induces Function Analysis: Blockade with a representative lot of anti-PD-L1 combined with IL-2 enhanced antiviral CD8 T cell responses during chronic LCMV infection (West, E.E., et. al. (2013). J Clin Invest. 123(6):2604-15).

Inhibition Analysis: A representative lot blocked the action of PD-L1 (Grabie, N., et. al. (2007). Circulation. 116(18):2062-71;West, E.E., et. al. (2013). J Clin Invest. 123(6):2604-15;El Annan, J., et. al. (2010). Invest Ophthalmol Vis Sci. 51(7):3418-23).

Quality

Evaluated by Flow Cytometry in EL4 cells.

Flow Cytometry Analysis: 1 µg/mL of this antibody detected PD-L1 in one million EL4 cells.

Target description

32.78 kDa calculated.

Linkage

Replaces: MABF405

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service