Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABC954

Sigma-Aldrich

Anti-MUTYH Antibody, clone 12D5.1

clone 12D5.1, from mouse

Synonym(s):

A/G-specific adenine DNA glycosylase, MutY homolog, hMYH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

12D5.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MUTYH(4595)

General description

A/G-specific adenine DNA glycosylase (UniProt Q9UIF7; also known as hMYH, MutY homolog) is encoded by the MUTYH (also known as MYH) gene (Gene ID 4595) in human. Base excision repair (BER) is initiated by damage-specific DNA glycosylases that recognize and remove aberrant bases by hydrolyzing the N-glycosidic bond, creating an apurinic/apyrimidinic (AP) site. This is followed by an (AP)-endonuclease-catalyzed DNA strand incision 5′ to the AP site, trimming of the 5′ end by an (AP)-lyase or an endonuclease, DNA resynthesis by a DNA polymerase and strand sealing by a DNA ligase. Monofunctional glycosylases have only glycosylase activity, whereas bifunctional glycosylases possess additional AP lyase activity and can therefore convert a base lesion into a single-strand break without the need for a separate AP endonuclease. MUTYH/MYH is a monofunctional DNA glycosylase that excises mismatched adenine opposite 7, 8-dihydro-8-oxoguanine (8-oxoG), which is the major mutagenic lesion induced by oxidative stress. Biallelic mutations in MUTYH are associated with MUTYH-Associated polyposis (MAP) and increased risk in colorectal cancer (CRC).

Specificity

Expected to react with all six spliced isoforms of human MUTYH reported by UniProt (Q9UIF7).

Immunogen

Epitope: N-terminal half.
GST-tagged recombinant internal fragment from the N-terminal half of human MUTYH.

Application

Anti-MUTYH Antibody, clone 12D5.1 is an antibody against MUTYH for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MUTYH in 10 µg of Raji cell lysate.

Target description

~59 kDa observed. 60.07 kDa (isoform Alpha-1), 59.14 kDa (isoform Alpha-2), 59.07 kDa (isoform Alpha-3), 58.44 kDa (isoform Beta-1), 57.52 kDa (isoform Gamma-2), and 50.52 kDa (isoform Gamma-3) calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service