Skip to Content
MilliporeSigma
All Photos(3)

Documents

MABC710

Sigma-Aldrich

Anti-TGFBR3, clone 1C5H11 Antibody

ascites fluid, clone 1C5H11, from mouse

Synonym(s):

Transforming growth factor beta receptor type 3, TGF-beta receptor type 3, TGFR-3, Betaglycan, Transforming growth factor beta receptor III, TGF-beta receptor type III, TGFBR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1C5H11, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
flow cytometry: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TGFBR3(7049)

Related Categories

General description

Transforming growth factor beta receptor type 3 (TGF-beta receptor type 3 or TGFR-3), also known as Betaglycan or Transforming growth factor beta receptor III (TGF-beta receptor type III), and encoded by the gene name TGFBR3, is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. TGFBR3 is extensively modified by glycosaminoglycan groups (GAG).

Immunogen

Purified recombinant fragment of human TGFBR3 expressed in E. Coli

Application

Anti-TGFBR3, clone 1C5H11 Antibody is an antibody against TGFBR3 for use in western blotting, flow cytometry & ELISA.
Flow Cytometry Analysis: A 1:200-1:400 dilution from a representative lot detected TGFBR3 in MOLT4 cells.

ELISA: A representative lot was used on Control Antigen (100 ng) and Antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).



Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers

Quality

Evaluated by Western Blotting in Jurkat, HeLa, MCF-7, F9, SK-N-SH and NIH/3T3 cell lysates.

Western Blotting Analysis: A 1:500-1:2,000 dilution of this antibody detected TGFBR3 in Jurkat, HeLa, MCF-7, F9, SK-N-SH and NIH/3T3 cell lysates.

Target description

~93 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service