Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC607

Sigma-Aldrich

Anti-MTSS1 Antibody, clone 15C3.2

clone 15C3.2, from mouse

Synonym(s):

Metastasis suppressor protein 1, Metastasis suppressor YGL-1, Missing in metastasis protein, MTSS1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$549.00

$549.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$549.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$549.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15C3.2, monoclonal

species reactivity

monkey, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTSS1(9788)

General description

Metastasis suppressor protein 1 (UniProt O43312; also known as Metastasis suppressor YGL-1, Missing in metastasis protein) is encoded by the MTSS1 (also known as KIAA0429, MIM) gene (Gene ID 9788) in human. MTSS1 is a scaffold protein that regulates cytoskeletal dynamics and actin polymerization. MTSS1 is also reported to bind Gli proteins or the Gli-Sufu complex and potentiate the transcription of effector genes of the sonic hedgehog signaling pathway. MTSS1 is down-regulated in many metastatic cancer cells compared with their non-metastatic counterparts. MTSS1 expression is shown to be inversely correlated with the prognosis of solid carcinomas of the breast, liver, and bladder.

Specificity

This clone is expected to react with all 4 spliced isoforms of human MTSS1.

Immunogen

Epitope: C-terminal
GST-tagged recombinant protein corresponding to the C-terminal of human MTSS1.

Application

Detect MTSS1 using this mouse monoclonal Anti-MTSS1 Antibody, clone 15C3.2, Cat. No. MABC607, validated for use in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected MTSS1 in human colon and human spleen tissues.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in COS-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MTSS1 in 10 µg of COS-1 cell lysate.

Target description

~82 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service