Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC590

Sigma-Aldrich

Anti-GRIM-19, clone 1A8 Antibody

clone 1A8, from mouse

Synonym(s):

NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13, Cell death regulatory protein GRIM-19, Complex I-B16.6, CI-B16.6, Gene associated with retinoic and interferon-induced mortality 19 protein, GRIM-19, Gene associated with retinoic and IFN-ind

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$622.00

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$622.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$622.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A8, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NDUFA13(51079)

General description

GRIM-19, also known as NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13, Cell death regulatory protein GRIM-19, Complex I-B16.6, CI-B16.6, Gene associated with retinoic and interferon-induced mortality 19 protein, Gene associated with retinoic and IFN-induced mortality 19 protein, NADH-ubiquinone oxidoreductase B16. GRIM-19 6 subunit, is encoded by the gene name NDUFA13, GRIM19, CDA016, CGI-39. GRIM-19 is accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I NADH dehydrogenase plays an important role in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. GRIM-19 is also suggested to be involved in the interferon/all-trans-retinoic acid (IFN/RA) induced cell death. This apoptotic activity is inhibited by interaction with viral IRF1. Additionally, GRIM-19 may play a role in CARD15-mediated innate mucosal responses and serve to regulate intestinal epithelial cell responses to microbes.GRIM-19 is expressed widely, with highest expression in heart, skeletal muscle, liver, kidney and placenta. In the intestinal mucosa, GRIM-19 is down-regulated in areas involved in Crohn disease and ulcerative colitis. Mutations in GRIM-19 have been associated with Hurthle cell thyroid carcinoma (HCTC), a rare type of thyroid cancer accounting for only about 3-10% of all differentiated thyroid cancers.

Immunogen

Recombinant protein corresponding to human GRIM-19.

Application

Anti-GRIM-19, clone 1A8 | MABC590 is an antibody against GRIM-19 for use in Western Blotting, Immunoprecipitation, Immunocytochemistry.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: A representative lot detected GRIM-19 in MCF-7 and RAW264.7 cell lysate (Hu, J., et al. (2002). J. Interferon & Cytokine Research. 22:1017-1026).
Immunoprecipitation Analysis: A representative lot immunoprecipitated GRIM-19 in MCF-7 cell lysate (Hu, J., et al. (2002). J. Interferon & Cytokine Research. 22:1017-1026).
Immunocytochemistry Analysis: A representative lot detected GRIM-19 in MCF-7 cells (Hu, J., et al. (2002). J. Interferon & Cytokine Research. 22:1017-1026).

Quality

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected GRIM-19 in 10 µg of MCF-7 cell lysate.

Target description

~16 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service