Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC537

Sigma-Aldrich

Anti-Vasohibin-1 Antibody, clone 4E12 (Azide Free)

clone 4E12, 1 mg/mL, from mouse

Synonym(s):

Vasohibin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$581.00

$581.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$581.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$581.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4E12, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

activity assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VASH1(22846)

General description

Vasohibin-1 is an endothelium-derived negative feedback regulator of angiogenesis. Vasohibin-1 inhibited migration, proliferation, and network formation by ECs in culture, as well as angiogenesis in vivo, and Vasohibin-1 was selective for endothelial cells and its influence was eliminated by hypoxia or TNF-alpha interaction. Vasohibin is preferentially expressed in endothelial cells and does not appear to promote proliferation or tumor growth when over expressed. Vasohibin-1 expression is induced by VEGF expression and its effects are mediated through VEGF-R2 independent of VEGF-R1 and Vasohibin-1 may directly influence the expression of VEGF-R2. Because the inhibition effects of Vasohibin-1 are nullified by hypoxia and various cytokines it is thought that this is how tumors escape this built-in negative regulator of angiogenesis. Vasohibin-1 is highly expressed in fetal organs and the endothelial cells during development, and also expressed in the brain and placental and microvessels endothelial cells of atherosclerotic lesions in the adult.

Immunogen

KLH-conjugated linear peptide corresponding to Human Vasohibin-1.

Application

Functional Activity Assay: A representative lot of this antibody induced cellular senescence (Miyashita, H., et al. (2012) PLOS ONE. 7(10):e46459).

Western Blotting Analysis: A representative lot of this antibody detected Vasohibin-1 in HUVECs transfected with control siRNA, but not in HUVECs transfected with Vasohibin-1 siRNA (Miyashita, H., et al. (2012) PLOS ONE. 7(10):e46459).

Western Blotting Analysis: A representative lot of this antibody detected Vasohibin-1 (KIAA1036 protein referenced in publication) in VEGF-stimulated HUVECs and GM7373 transfected cell lysate (Watanabe, K,, et al. (2004) JCI. 114(7):898-907).

Immunohistochemistry Analysis: A representative lot of this antibody detected Vasohibin-1 (KIAA1036 protein referenced in publication) in human placenta tissue (Watanabe, K,, et al. (2004) JCI. 114(7):898-907).

Immunocytochemistry Analysis: A representative lot of this antibody detected Vasohibin-1 (KIAA1036 protein referenced in publication) in HUVEC cells (Watanabe, K,, et al. (2004) JCI. 114(7):898-907).
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
This Anti-Vasohibin-1 Antibody, clone 4E12 (Azide Free) is validated for use in Western Blotting and Immunohistochemistry and Immunocytochemistry and Activity Assay for the detection of Vasohibin-1.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Vasohibin-1 in 10 µg of HEK293 cell lysate.

Target description

~ 41 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siem van der Laan et al.
Cell reports, 29(12), 4159-4171 (2019-12-19)
The two related members of the vasohibin family, VASH1 and VASH2, encode human tubulin detyrosinases. Here we demonstrate that, in contrast to VASH1, which requires binding of small vasohibin binding protein (SVBP), VASH2 has autonomous tubulin detyrosinating activity. Moreover, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service