Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC307

Sigma-Aldrich

Anti-phospho-ULK1 (Ser 467), clone 15D12.3 Antibody

clone 15D12.3, from mouse

Synonym(s):

Serine/threonine-protein kinase ULK1, Autophagy-related protein 1 homolog, ATG1, hATG1, Unc-51-like kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$566.00

$566.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$566.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$566.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15D12.3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgMλ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer467)

Gene Information

human ... ULK1(8408)

General description

ULK1 or autophagy-related protein 1 homolog (ATG1) is a serine/threonine protein kinase. It is a ubiquitously expressed protein that is highly abundant in the brain, skeletal muscle, and heart. It is involved in the signaling pathway of mTOR receptors leading to the regulation of autophagy in nutrient-deprived cells. mTOR may interact with ULK1 as a complex of ULK1/FIP200/Atg13. ULK1 may play an important role in neuronal development: Elimination of ULK1′s enzymatic activity by targeted disruption of its kinase domain has blocked the outgrowth of neurons in the developing mouse brain, possibly by blocking the ULK1-mediated trafficking of neurotrophins.

Immunogen

KLH-conjugated linear peptide corresponding to human phospho-ULK1 (Ser 467).

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-phospho-ULK1 (Ser 467) antibody, clone 15D12.3 is validated for use in western blotting for the detection of phospho-ULK1 (Ser 467).

Quality

Evaluated by Western Blotting in A-431 serum starved cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected phospho-ULK1 (Ser467) in 10 µg of A-431 serum starved cell lysate.

Target description

~112 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgMλ supernatant in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service