Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1786

Sigma-Aldrich

Anti-Glypican-2 Antibody, clone CT3

Synonym(s):

GPC2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$195.00
100 μL
$491.00

$195.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
$195.00
100 μL
$491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$195.00


Estimated to ship onMarch 31, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CT3, monoclonal

mol wt

calculated mol wt 62.83 kDa
observed mol wt ~56 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... GPC2(221914)

Specificity

Clone CT3 is a mouse monoclonal antibody that detects Glypican-2 (GPC2) and its glycated forms. It targets an epitope within the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 50 amino acids from the C-terminal region of human Glypican-2 (GPC2).

Application

Quality Control Testing

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Glypican-2 in SH-SY5Y cell lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected Glypican-2 in IMR-32 cell lysate.

Immunohistochemistry Applications: A representative lot detected Glypican-2 in Immunohistochemistry applications (Li, N., et al. (2021). Cell Rep Med. 2(6): 100297; Li, N., et al. (2020). J Histochem Cytochem. 68(12): 841-862).

ELISA: A representative lot detected Glypican-2 in ELISA applications (Li, N., et al. (2021). Cell Rep Med. 2(6): 100297).

Flow Cytometry Analysis: A representative lot detected Glypican-2 in Flow Cytometry applications (Li, N., et al. (2021). Cell Rep Med. 2(6): 100297; Li, N., et al. (2020). J Histochem Cytochem. 68(12): 841-862).

Western Blotting Analysis: A representative lot detected Glypican-2 in Western Blotting applications (Li, N., et al. (2021). Cell Rep Med. 2(6):100297).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Glypican-2 (UniProt: Q8N158; also known as GPC2) is encoded by the GPC2 gene (Gene ID: 221914) in human. Glypicans belong to a family of heparan sulfate proteoglycans that are attached to the cell membrane via a GPI-anchor. They interact with multiple ligands, including morphogens, growth factors, chemokines, ligands, receptors, and components of the extracellular matrix through their heparan sulfate chains and core protein. GPC2 is synthesized with a signal sequence (aa 1-23), which is subsequently cleaved off to produce the mature GPC2 (aa 24-554). It also contains a propeptide sequence (aa 555-579) that is removed in the mature form. Glypicans are reported to regulate Wnt/β-catenin signaling and affect cell growth and proliferation. Overexpression of GPC2 is reported in several pediatric cancers. Its higher expression is also shown in neuroblastoma compared to the normal peripheral nerve tissue. GPC2 can modulate Wnt signaling and regulate MYCN in neuroblastoma pathogenesis. Genetically silencing of CPC2 is shown to inhibit neuroblastoma cell growth by suppressing N-Myc expression. Clone CT3 specifically binds to GPC2 in human cancer cells and does not react with normal tissues. It recognizes tumor-associated exons 3 and 10 of GPC2. (Ref.: Li, N., et al. (2020). J. Histochem. Cytochem. 68(12); 841-862; Li, N., et al., 2021, Cell Rep. Medicine 2, 100297).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service