Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABC166

Sigma-Aldrich

Anti-Angiogenin Antibody, clone 26–2F (Azide Free)

clone 26-2F, from mouse

Synonym(s):

Angiogenin, Ribonuclease 5, RNase 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

26-2F, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANG(283)

General description

Angiogenin (Ang) is a non-glycosylated polypeptide, 123 amino acids in length, with a molecular mass of 14 kDa. It is secreted by tumor cells and is a potent inducer of neovascularization. Angiogenin is produced by a variety of cell types including vascular endothelial cells and smooth muscle cells, fibroblasts, tumor colonic epithelium, normal peripheral blood lymphocytes, lung and colonic epithelial tumor cell lines, and primary gastrointestinal adenocarcinomas. It specifically binds to endothelial cells and elicits second messenger systems. Angiogenin is present in human follicular fluid, and its production is up-regulated by human chorionic gonadotropin and hypoxic conditions. Angiogenin shows a high degree of homology with known ribonucleases such as pancreatic ribonuclease A, and the capacity of angiogenin to induce blood vessel growth is dependent on its ribo-nucleolytic activity. Angiogenin is thought to be involved in the development of solid tumors; angiogenin antagonists are capable of inhibiting tumor growth. Angiogenin is endocytosed by subconfluent endothelial cells and translocated to the nucleus, where it accumulates in the nucleolus. Serum angiogenin concentration is increased in serum of colorectal cancer patients, and correlates with cancer progression. The cytoskeletal protein α-actinin-2 binds to angiogenin, which might be a novel interface target for anti-angiogenesis and anti-tumor therapy.

Immunogen

Purified Human Angiogenin.

Application

Inhibition Assay: A representative lot of this antibody blocked cell-free protein synthesis by angiogenin (Fett et al., (1994) Biochemistry. 33(18):5421-5427).

Inhibition Assay: A representative lot of this antibody inhibited ribonucleolytic activity (Piccoli et al., (1998) Proc. Natl. Acad. Sci. 95:4579–4583).

Immunohistochemistry Analysis: A representative lot of this antibody detected Angiogenin in Neovessels of cancer tissue (Tsuji et al., (2005) Cancer Res. 65:1352-1360).

Immunocytochemistry Analysis: A representative lot of this antibody detected Angiogenin in HeLa and Huvec cells (Tsuji et al., (2005) Cancer Res. 65:1352-1360).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Angiogenin Antibody, clone 26–2F (Azide Free) is validated for use in Immunocytochemistry and Inhibition and Immunohistochemistry for the detection of Angiogenin.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 4 µg/mL of this antibody detected Angiogenin in Angiogenin-treated and untreated HeLa Cells. HeLa cells that were treated with Angiogenin up-took the protein and translocated it to the nucleus. Untreated HeLa cells did not exhibit any Angiogenin protein signal.

Target description

16 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksej Drino et al.
Nucleic acids research, 51(3), 1326-1352 (2023-02-01)
Stress-induced tRNA fragmentation upon environmental insult is a conserved cellular process catalysed by endonucleolytic activities targeting mature tRNAs. The resulting tRNA-derived small RNAs (tsRNAs) have been implicated in various biological processes that impact cell-to-cell signalling, cell survival as well as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service