Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC1211

Sigma-Aldrich

Anti-Sorting Nexin-17 Antibody, clone 4B8.1

clone 4B8.1, from mouse

Synonym(s):

Sorting nexin-17, SNX17

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$560.00

$560.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$560.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$560.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4B8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SNX17(9784)

General description

Sorting nexin-17 (UniProt Q15036) is encoded by the SNX17 (also known as KIAA0064) gene (Gene ID 9784) in human. The sorting nexins (SNXs) constitute a family of adaptor proteins involved in proteins trafficking through the endosomal pathway. Originally identified as a binding partner of the cell adhesion protein P-selectin, sorting nexin-17 (SNX17) has since been shown to mediate internalization and recycling of multiple cell surface proteins including P-selectin, amyloid precursor protein (APP), integrin β1, low density lipoprotein receptor (LDLR), LDLR related protein (Lrp1), ApoER2, and FEEL1. All SNX proteins contain a conserved phox homology (PX) domain that mediates phosphoinositides binding in a SNX type-specific manner, as well as various scaffolding modules including SH3, BAR, and PDZ domains. SNX17 PX domain (a.a. 1-109) displays specificity for phosphatidylinositol 3-monophosphate, which is enriched at the cytoplasmic face of the early endosome. SNX17 also contains a 4.1 protein/ezrin/radixin/moesin (FERM) domain (a.a. 115-432) comprised of an ubiquitin/Ras association (RA) subdomain known as the F1 lobe (a.a. 113-205), an F2 lobe (a.a. 206-269) with a helical structure similar to acyl-CoA binding protein, and an F3 lobe (a.a. 270-388) similar to the phosphotyrosine binding (PTB)/pleckstrin homology domain. The FERM domain mediates interaction with Asn-Pro-Xaa-Tyr/Phe (NPX(Y/F) motif-containing proteins for internalization as well as interaction with adaptor proteins-containing PTB (or FERM) domains to trigger signaling events.

Specificity

Clone 4B8.1 recognizes an internal epitope within the FERM-like region present in both spliced isoforms of human sorting nexin-17 reported by UniProt (Q15036).

Immunogen

Epitope: Within the FERM-like region.
GST-tagged recombinant fragment from the FERM-like region of human Sorting Nexin-17.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Sorting Nexin-17 in human pancreas, prostate, and cerebral cortex tissue sections.
Research Category
Apoptosis & Cancer
Research Sub Category
Vesicular Trafficking
This Anti-Sorting Nexin-17 Antibody, clone 4B8.1 is validated for use in Western Blotting, Immunohistochemistry for the detection of Sorting Nexin-17.

Quality

Evaluated by Western Blotting in U251 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Sorting Nexin-17 in 10 µg of U251 cell lysate.

Target description

~52 kDa observed. 52.90 kDa (isoform 1) and 50.00 kDa (isoform 2) calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service