Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC1190

Sigma-Aldrich

Anti-Bin1 Antibody, clone 2F11

clone 2F11, from mouse

Synonym(s):

Myc box-dependent-interacting protein 1, Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$546.00

$231.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1067

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$546.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1067

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F11, monoclonal

species reactivity

mouse, rat, chicken, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ
IgG1

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

chicken ... Bin1(424228)
human ... BIN1(274)
mouse ... Bin1(30948)

General description

Myc box-dependent-interacting protein 1 (UniProt: O00499; also known as Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1) is encoded by the BIN1 (also known as AMPHL) gene (Gene ID: 274) in human. Bin1 is a ubiquitous protein that displays highest expression levels in the brain and muscle. Eleven isoforms of Bin1 have been described that are generated by alternate splicing. Isoform IIA is reported to be expressed only in the brain where it is concentrated n axon initial segments and nodes of Ranvier. It contains one BAR domain (aa 29-276) and one SH3 domain (aa 520-592). Bin1 interacts with the functionally critical Myc box regions at the N- terminus of the MYC oncoprotein and inhibits MYC-induced malignant cell transformation. Hence, it is considered as a tumor suppressor. Mutations in BIN1 gene are known to cause centronuclear myopathy that is characterized by progressive muscular weakness and wasting involving mainly limb girdle, trunk, and neck muscles.

Specificity

Clone 2F11 recognizes an epitope within the N-terminal BAR domain of Bin 1.

Immunogen

Epitope: domain of:
GST-tagged recombinant fragment corresponding to the BAR domain (aa 29-276) from the N-terminal half of human Myc box-dependent-interacting protein 1 (Bin1).

Application

Anti-Bin1, clone 2F11 Antibody, Cat. No. MABC1190, is a mouse monoclonal antibody that detects Bridging integrator 1 (Bin1) protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Detect Bin1 using this mouse monoclonal Anti-Bin1, clone 2F11 Antibody, Cat. No. MABC1190-25UG, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: 4 µg/mL from a representative lot detected Bin1 in 10 µg of LNCaP and C2C12 cell lysates.
Immunohistochemistry Analysis: A representative lot detected Bin1 in Immunohistochemistry applications (DuHadaway, J.B., et. al. (2003). J Cell Biochem. 88(3):635-42).
Western Blotting Analysis: A representative lot detected Bin1 in Western Blotting applications (DuHadaway, J.B., et. al. (2003). J Cell Biochem. 88(3):635-42; Muller, A.J., et. al. (2003). Mol Cell Biol. 23(12):4295-306; Chang, M.Y., et. al. (2007). Cancer Res. 67(1):100-7).
Western Blotting Analysis: 4 µg/mL from a representative lot detected Bin1 in 10 µg of LNCaP and C2C12 cell lysates.

Immunohistochemistry Analysis: A representative lot detected Bin1 in Immunohistochemistry applications (DuHadaway, J.B., et. al. (2003). J Cell Biochem. 88(3):635-42).

Western Blotting Analysis: A representative lot detected Bin1 in Western Blotting applications (DuHadaway, J.B., et. al. (2003). J Cell Biochem. 88(3):635-42; Muller, A.J., et. al. (2003). Mol Cell Biol. 23(12):4295-306; Chang, M.Y., et. al. (2007). Cancer Res. 67(1):100-7).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human cerebellum and human skeletal muscle tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected Bin1 in human cerebellum and human skeletal muscle tissue.

Target description

64.70 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service