Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1170

Sigma-Aldrich

Anti-MNDA Antibody, clone 3C1

clone 3C1, from rat

Synonym(s):

Myeloid cell nuclear differentiation antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C1, monoclonal

species reactivity

human

technique(s)

dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MNDA(4332)

Related Categories

General description

Myeloid cell nuclear differentiation antigen (UniProt P41218) is encoded by the MNDA gene (Gene ID 4332) in human. MNDA is a member of the interferon-inducible p200 (IFI-200) family of proteins with a conserved HIN-200 domain (human MNDA a.a. 196-394) that mediate interaction with proteins and DNA. IFI-200 members possess antimicrobial, cell growth regulatory, differential regulatory and immunomodulatory properties. In bone marrow-derived- and mature neutrophils, MNDA is predominantly located in the nucleus. MNDA is cleaved by caspases in neutrophils undergoing apoptosis and relocated to the cytoplasm. MNDA is downregulated in myelodysplastic syndrome and completely absent in a mouse strain susceptible to B-cell plasmacytoma, while being markedly (1,000-fold) upregulated in a mouse strain resistant to B-cell plasmacytoma. Originally thought to be restricted to, myeloid cells, MNDA and other IFI-200 family members are also found to be expressed in a variety of non-myeloid cell types. MNDA is highly expressed in normal bone tissue, while being at markedly low levels in osteosarcoma cells. MNDA overexpression is shown to induce apoptosis, inhibit proliferation, and reduce the migration of osteosarcoma cells.

Specificity

Clone 3C1 reacted with both denatured and non-denatured MNDA. Clone 3C1 reacted with nuclear extract from NMDA-expressing HL-60, but not non-MNDA-expressing BII, human myeloid leukemia cells (Hudson, C.R., et al. (1988). Hybridoma. 7(6):541-553).

Immunogen

Human peripheral blood granulocyte nuclei.

Application

Detect MNDA using this rat monoclonal Anti-MNDA Antibody, clone 3C1, Cat. No. MABC1170, validated for use in Dot Blot, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Dot Blot Analysis: Clone 3C1 hybridoma culture supernatant reacted with nuclear extract from HL-60, but not BII, human myeloid leukemia cells (Hudson, C.R., et al. (1988). Hybridoma. 7(6):541-553).

Flow Cytometry Analysis: A representative lot (pre-conjugated with Alexa Fluor 488) detected MNDA expression among the granulocyte-macrophage (G-M) progenitors, but not among the nucleated red blood cells (nRBCs) and lymphoid cells in control and myelodysplastic syndromes (MDS) human marrow aspirates. A G-M progenitor cell population with downregulated MNDA expression was detected in most marrow samples from MDS cases and some MDS cases contain MNDA-negative G-M progenitor cells (Briggs, R.C., et al. (2006). Cancer Res. 66(9):4645-4651).

Immunocytochemistry Analysis: A representative lot detected MNDA nuclear immunoreactivity among HL-60 cells and NMDA-transfected, but not untransfected K562 cells. MNDA-negative HL-60 cells increased with time in cultures maintained without fresh medium for an extended period of time. MNDA translocated from nucleus to cytoplasm beginning at 3 hours and completed 4 hours following H2O2 addition to HL-60 cells (Briggs, R.C., et al. (2006). Cancer Res. 66(9):4645-4651).

Immunoprecipitation Analysis: A representative lot immunoprecipitated MNDA from HL-60 nuclear extract (Hudson, C.R., et al. (1988). Hybridoma. 7(6):541-553).

Western Blotting Analysis: A representative lot detected a time-dependent loss of MNDA in HL-60 cell lysate beginning at 4 hours following H2O2 treatment (Briggs, R.C., et al. (2006). Cancer Res. 66(9):4645-4651).

Western Blotting Analysis: Clone 3C1 hybridoma culture supernatant detected MNDA in HL-60 nuclear extract (Hudson, C.R., et al. (1988). Hybridoma. 7(6):541-553).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunocytochemistry in HL-60 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution of this antibody detected MNDA in 4% paraformaldehyde-fixed, 0.3% Triton X-100-permeabilized HL-60 cells.

Target description

45.84 kDa calculated. ~55 kDa reported (Hudson, C.R., et al. (1988). Hybridoma. 7(6):541-553). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefania Bottardi et al.
Experimental hematology, 88, 68-82 (2020-07-19)
The myeloid nuclear differentiation antigen (MNDA) is a stress-induced protein that promotes degradation of the anti-apoptotic factor MCL-1 and apoptosis in myeloid cells. MNDA is also expressed in normal lymphoid cells and in B-cell clones isolated from individuals with chronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service