Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC1151

Sigma-Aldrich

Anti-NY-ESO-1 Antibody, clone D8.38

clone D8.38, from mouse

Synonym(s):

Cancer/testis antigen 1, Autoimmunogenic cancer/testis antigen NY-ESO-1, Cancer/testis antigen 6.1, CT6.1, L antigen family member 2, LAGE-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$567.00

$567.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1540


Select a Size

Change View
100 μL
$567.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$567.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1540

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

D8.38, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CTAG1A(246100)

General description

Cancer/testis antigen 1 (UniProt: P78358; also known as Autoimmunogenic cancer/testis antigen NY-ESO-1, Cancer/testis antigen 6.1, CT6.1, L antigen family member 2, LAGE-2) is encoded by the CTAG1A (also known as CTAG, CTAG1, ESO1, LAGE2, LAGE2A, CTAG1B, LAGE2B) gene (Gene ID: 1485; 246100) in human. NY-ESO-1 is a human tumor antigen of the cancer/testis family that is highly expressed in many poor-prognosis melanomas and in other cancer types. However, it is not detected in non-malignant tissues with the exception of germ cells and trophoblasts. It contains a C-terminal hydrophobic tail that potentially serves as membrane-associated domain. NY-ESO-1 is expressed from 18 weeks until birth in human fetal testis and in adult testis it is strongly expressed in spermatogonia and in primary spermatocytes, but not in post-meiotic cells or in testicular somatic cells. Although the exact function of NY-ESO-1 is still not clear, it has been speculated it might contribute to certain properties, such as immortality, self-renewal, migratory ability, and capacity to invade that favor tumor cell survival. On the basis of its immunogenicity, NY-ESO-1 is considered as one of the most attractive antigens for cancer immunotherapy. T cells engineered to express an affinity-enhanced T cell receptor (TCR) recognizing a naturally processed peptide shared by the cancer-testis antigens NY-ESO-1 and LAGE-1 have been considered as an attractive method for treatment of myeloma and enhance long-term survival of cells.

Specificity

Clone D8.38 specifically targets a sequence in the N-terminal region and detects Cancer/testis antigen 1 (NY-ESO1) in human testis

Immunogen

A full length recombinant human NY-ESO-1 protein.
Epitope: domain of:

Application

Detect NY-ESO-1, LAGE-1 using this mouse monoclonal Anti-NY-ESO-1 antibody, clone D8.38, Cat. No. MABC1151. Validated for use in Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected NY-ESO-1 in human testis tissue.

Immunohistochemistry Analysis: A representative lot detected NY-ESO-1 in Immunohistochemistry applications. (Yakirevich, E., et. al. (2003). Clin Cancer Res. 9(17):6453-60; Bujas, T., et. al. (2011). Eur J Histochem. 55(1):e7; Bolli, M., et. al. (2005). Int J Cancer. 115(6):960-6; Groeper, C., et. al. (2007). Int J Cancer. 120(2):337-43).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry in human testis tissue.

Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected NY-ESO-1 in human testis tissue.

Target description

17.99 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martine Bloemendal et al.
Oncoimmunology, 11(1), 2015113-2015113 (2022-12-17)
We evaluated the immunological responses of lymph-node involved (stage III) melanoma patients to adjuvant dendritic cell vaccination with subsets of naturally occurring dendritic cells (nDCs). Fifteen patients with completely resected stage III melanoma were randomized to receive adjuvant dendritic cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service