Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC1138

Sigma-Aldrich

Anti-HPCAL1 Antibody, clone 5A5

clone 5A5, from mouse

Synonym(s):

Hippocalcin-like protein 1, Calcium-binding protein BDR-1, HLP2, Visinin-like protein 3, VILIP-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A5, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... HPCAL1(3241)

General description

Hippocalcin-like protein 1 (UniProt: P37235; also known as Calcium-binding protein BDR-1, HLP2, Visinin-like protein 3, VILIP-3) is encoded by the HPCAL1 (also known a BDR1) gene (Gene ID: 3241) in human. HPCAL1 is a lipid-anchor membrane protein of the visinin-like subfamily of neural calcium sensor and is involved in the calcium-dependent regulation of rhodopsin phosphorylation. It is structurally related to Ca2+-myristoyl switch proteins that contain four EF-hand motifs and a covalently attached N-terminal myristoyl group. HPCAL1 is also expressed in Purkinje cells of the cerebellum and hippocampus where it is believed to regulate synaptic plasticity relevant for learning and memory. Clone 5A5 can detect HPCAL1 in formalin-fixed 293 cells and in pancreatic cell lines. (Ref.: Chen, Y., et al. (2014). Monoclonal Antibodies in Immunodiagnosis and Immunotherapy 33(1); 20-27; Li, C., et al. (2016). PLoS One 11(11); e0165921).

Specificity

Clone 5A5 specifically detects Hippocalcin-like protein 1 (HPCAL1) in human cells.

Immunogen

Native and denatured human HPCAL1.

Application

Detects Hippocalcin-like protein 1 using this mouse monoclonal Anti-HPCAL1, clone 5A5, Cat. No. MABC1138, tested for use in Western Blotting, Immunohistochemistry (Paraffin), ELISA and Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected HPCAL1 in Immunocytochemistry applications (Chen, Y., et. al. (2014). Monoclon Antib Immunodiagn Immunother. 33(1):20-7).

ELISA Analysis: A representative lot detected HPCAL1 in ELISA applications (Chen, Y., et. al. (2014). Monoclon Antib Immunodiagn Immunother. 33(1):20-7).

Immunohistochemistry (Paraffin) Analysis: A 1:50-250 Dilution from a representative lot detected HPCAL1 in human kidney and human cerebellum tissue sections.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in Panc-1 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected HPCAL1 in Panc-1 cell lysate.

Target description

~22 kDa observed; 22.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service