Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC112

Sigma-Aldrich

Anti-RASSF1 Antibody, clone 5C9.1

clone 5C9.1, from mouse

Synonym(s):

Ras association domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RASSF1(11186)

General description

RASSF1, also known as Ras association domain-containing protein 1, and encoded by the gene RASSF1/RDA32, is an important cell cycle and cell proliferation regulator protein. RASSF1 acts as a tumor suppressor and operates within the death receptor dependent apoptotic pathways. Additionally, RASSF1 inhibits proliferation at the G1/S boundary by regulating the accumulation of cyclin D1 protein. RASSF1 is localized to the cytoplasm and cytoskeleton depending upon cellular environment. RASSF1 localizes to cytoplasmic microtubules during interphase, to bipolar centrosomes associated with microtubules during prophase, to spindle fibers and spindle poles at metaphase and anaphase, to the midzone during early telophase, and to the midbody in late telophase in cells. The protein also co-localizes with MDM2 in the nucleus. RASSF1 is expressed in multiple isoforms but in generally it is ubiquitously expressed but down regulated in cancer and tumor cells.

Immunogen

GST-tagged recombinant protein corresponding to human RASSF1.

Application

Detect RASSF1 using this Anti-RASSF1 Antibody, clone 5C9.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50-500 dilution from a representative lot detected RASSF1 in human kidney and human prostate cancer tissue.

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected RASSF1 in 10 µg of human brain tissue lysate.

Target description

~39 kDa observed

Linkage

Replaces: MABE526

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service