Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABC1022

Sigma-Aldrich

Anti-BCL2L12 Antibody, clone 11B5.1

clone 11B5.1, from mouse

Synonym(s):

Bcl-2-like protein 12

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$570.00

$570.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$570.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$570.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11B5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BCL2L12(83596)

General description

Bcl-2-like protein 12 (UniProt Q9HB09; also known as Bcl-2-related proline-rich protein, Bcl2-L-12) is encoded by the BCL2L12 (also known as BPR) gene (Gene ID 83596) in human. Bcl-2 family proteins paly either a pro- or anti-apoptotic role in cell death signaling. BCL-2 family members are related by regions of sequence and structural homology. The pro-survival members contain four BCL-2 homology regions (BH1 BH4), whereas the BH3-only members contain only the BH3 amphipathic helix that mediates their interaction with the groove of multi-domain BCL-2 family members. Bcl2-L-12 is an anti-apoptotic protein that shares a lower amino acid similarity with the other Bcl2 family members. It contains a BH2 (a.a. 311-322) domain and a BH3-like domain (a.a. 192-220) that mediate its interaction with Bcl2 and Bcl-xL. Exogenous expression of Bcl2-L-12 with Leu213 or Leu217 mutation in U87MG cell line is reported to result in reactivation of PARP cleavage, caspase-3 and cytochrome c release compared to cells expressing wild-type Bcl2-L-12.

Specificity

Clone 11B5.1 targets an internal epitope present in spliced isoforms 1&3, but absent in isoform 2, of human BCL2L12 reported by UniProt (Q9HB09).

Immunogen

GST-tagged recombinant human BCL2L12 internal fragment.

Application

Detect Bcl2L12 using this mouse monoclonal Anti-BCL2L12, clone 11B5.1 Antibody, Cat. No. MABC1022, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BCL2L12 in human pancreas, skeletal muscle, and tonsil tissue sections.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected BCL2L12 in 10 µg of PC3 human prostate cancer cell lysate.

Target description

~37 kDa observed. 36.82/36.67 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service