Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC006F

Sigma-Aldrich

Mouse IgG2b Negative Control, clone GC198, FITC conjugate

Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$258.00

$258.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$258.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$258.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

clone

GC198, monoclonal

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

Specificity

This antibody reacts with the outer membrane protein of some strains of Neisseria gonorrhoeae. No reaction with human cell surface or plasma components has been observed. However, some binding to Fc receptors may occur.

Immunogen

Protein I preparation from Neisseria gonorrhoeae.

Application

FITC conjugated mouse IgG2b is a monoclonal antibody intended for use as a negative control in flow cytometry. Its use enables an estimation of non-specific binding of mouse monoclonal antibodies to cell surface components in peripheral blood and tissue.

SUGGESTED USAGE

Flow cytometry - use 10 μl direct from the vial per 100 μl of whole blood, or 1 x 10E6 peripheral blood mononuclear cells (PBMC) in 100 μl buffer.
Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.
Research Category
Secondary & Control Antibodies

Epitope Tags & General Use
Research Sub Category
Isotype Control Antibodies

Physical form

Purified immunoglobulin. Fluorescein conjugated, ready to use. The antibody is supplied in 1.0ml phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.

Storage and Stability

Store at 2 to 8oC, for up to 6 months. DO NOT FREEZE. Protect from light.

WARNING

The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Narges Labibzadeh et al.
Cell journal, 18(3), 302-309 (2016-09-08)
Nonunion is defined as a minimum of 9 months since injury without any visible progressive signs of healing for 3 months. Recent literature has shown that the application of mesenchymal stromal cells is safe, in vitro and in vivo, for
Mohsen Emadedin et al.
Cell journal, 19(1), 159-165 (2017-04-04)
Nonunion is defined as a minimum of a 9-month period of time since an injury with no visibly progressive signs of healing for 3 months. Recent studies show that application of mesenchymal stromal cells (MSCs) in the laboratory setting is
Leila Satarian et al.
Journal of ophthalmic & vision research, 12(1), 58-64 (2017-03-17)
To examine the safety of a single intravitreal injection of autologous bone Marrow Mesenchymal stem cells (MSCs) in patients with advanced retinitis pigmentosa (RP). A prospective, phase I, nonrandomized, open-label study was conducted on 3 eyes of 3 volunteers with
Fattah Sotoodehnejadnematalahi et al.
Cell journal, 23(1), 1-13 (2021-03-03)
In the present study, we examined the tolerance-inducing effects of human adipose-derived mesenchymal stem cells (hAD-MSCs) and bone marrow-derived MSCs (hBM-MSCs) on a nonhuman primate model of skin transplantation. In this experimental study, allogenic and xenogeneic of immunomodulatory properties of
Ni Sima et al.
Orphanet journal of rare diseases, 13(1), 54-54 (2018-04-11)
Infantile and late infantile neuronal ceroid lipofuscinoses (NCLs) are lysosomal storage diseases affecting the central nervous system (CNS). The infantile NCL (INCL) is caused by mutations in the PPT1 gene and late-infantile NCL (LINCL) is due to mutations in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service