Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB945

Sigma-Aldrich

Anti-Coxsackievirus B6 Antibody, clone 363-9E-2F-11E

clone 363-9E-2F-11E, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

363-9E-2F-11E, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with Coxsackie virus B6. Neutralizes Coxsackie virus B6 Schmitt strain at a titer of less than 1:16.

Application

Anti-Coxsackievirus B6 Antibody, clone 363-9E-2F-11E is an antibody against Coxsackievirus B6 for use in IF.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
Suggested starting dilution for IFA is 1:1000.

Dilute with buffer pH 7.5 -8.0 containing protein or other stabilizing medium. Final working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PB with 0.25M NaCl, pH=7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service