Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB8716RU

Sigma-Aldrich

Anti-Rabies Antibody, clone C4-62-15-2

isotype IgG1, purified antibody, suitable for immunofluorescence

Synonym(s):

Rabies Antiserum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C4-62-15-2, monoclonal

purified by

using Protein A (Affinity Purified)

species reactivity

rabies virus

packaging

antibody small pack of 100 μg

technique(s)

immunofluorescence: suitable

isotype

IgG1

shipped in

2-8°C

Immunogen

Rabies virus

Application

Quality Control Testing

This mouse monoclonal Anti-Rabies clone C4-62-15-2, Cat. No. MAB8716RU, is tested for use in Immunofluorescence, for the detection of Rabies Virus.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified mouse monoclonal antibody in 0.02 M phosphate buffer, 0.25 M NaCl, pH 7.6, containing 0.1% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service