Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB8257B-5

Sigma-Aldrich

Anti-Influenza A Antibody, nucleoprotein, clone A1

clone A1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$1,900.00

$1,900.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

Select a Size

Change View
500 μG
$1,900.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$1,900.00


Please contact Customer Service for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1020

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Specific for the Influenza A nucleoprotein. Has stronger binding with N1-type Flu A. Has been shown to react with the H5N1 strain. No cross reactivity seen to influenza B or other respiratory viruses.

Immunogen

Epitope: nucleoprotein
Influenza A

Application

Anti-Influenza A Antibody, nucleoprotein, clone A1 is an antibody against Influenza A for use in IF.
Immunofluorescence

Optimal dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Biotin conjugated purified immunoglobulin. Liquid in 0.01M PBS, pH=7.1, 0.1% Sodium Azide with 15 mg/mL BSA as stabilizer.

Storage and Stability

Maintain at 2 to 8°C for up to 12 months from date of receipt. Protect from Light.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibodies for the rapid diagnosis of influenza A and B virus infections by immunofluorescence.
McQuillin, J, et al.
Lancet, 2, 911-914 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service