Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB80132

Sigma-Aldrich

Anti-Human Metapneumovirus Antibody, clone 406

clone 406, Chemicon®, from mouse

Synonym(s):

hMPV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

406, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

General description

Human metapneumovirus (hMPV) is a member of the pneumovirinae subfamily of paramyxoviruses, first described in 2001 from pediatric respiratory specimens in the Netherlands. It is an enveloped pleomorphic virus with a single negative-strand RNA genome. Two major serotypes, A and B, have been described. Several studies identified hMPV in specimens worldwide, and estimated that by age 5 years 70% of children have developed antibodies to hMPV.

Specificity

Reactive with sub-types A1, A2, B1, and B2. Specific to the nuclear protein.

Application

Anti-Human Metapneumovirus Antibody, clone 406 detects level of Human Metapneumovirus & has been published & validated for use in ELISA, FC.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service