Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB80122

Sigma-Aldrich

Anti-Human Metapneumovirus Antibody, clone 67

clone 67, Chemicon®, from mouse

Synonym(s):

hMPV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$557.00

$557.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$557.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

$557.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

67, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

Related Categories

General description

Human metapneumovirus (hMPV) is a member of the pneumovirinae subfamily of paramyxoviruses, first described in 2001 from pediatric respiratory specimens in the Netherlands. It is an enveloped pleomorphic virus with a single negative-strand RNA genome. Two major serotypes, A and B, have been described. Several studies identified hMPV in specimens worldwide, and estimated that by age 5 years 70% of children have developed antibodies to hMPV.

Specificity

Specific to the fusion protein. Reactive with all sub-types; A1, A2, B1, B2.

Application

Detect Human Metapneumovirus using this Anti-Human Metapneumovirus Antibody, clone 67 validated for use in ELISA, FC & IF.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.

Storage and Stability

Store at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles J Russell et al.
Virology, 509, 60-66 (2017-06-13)
Human metapneumovirus (hMPV) infections pose a serious health risk to young children, particularly in cases of premature birth. No licensed vaccine exists and there is no standard treatment for hMPV infections apart from supportive hospital care. We describe the production
Analysis of the genomic sequence of a human metapneumovirus
van den Hoogen, Bernadette G, et al
Virology, 295, 119-132 (2002)
Human metapneumovirus associated with respiratory tract infections in a 3-year study of nasal swabs from infants in Italy.
Maggi, Fabrizio, et al.
Journal of Clinical Microbiology, 41, 2987-2991 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service