Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB5620

Sigma-Aldrich

Anti-Norepinephrine Transporter Antibody

ascites fluid, Chemicon®

Synonym(s):

NET

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$699.00

$699.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$699.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$699.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Norepinephrine Transporter, human and non-human primate.

Immunogen

Synthetic peptide from human norepinephrine Transporter.

Application

Detect Norepinephrine Transporter using this Anti-Norepinephrine Transporter Antibody validated for use in IH.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Neukirchen et al.
British journal of anaesthesia, 109(6), 887-896 (2012-09-05)
Intraoperative hypotension is associated with increased risk of perioperative complications. The N-methyl-d-aspartate (NMDA) receptor (NMDA-R) antagonist xenon (Xe) induces general anaesthesia without impairment of cardiac output and vascular resistance. Mechanisms involved in cardiovascular stability have not been identified. Muscle sympathetic
Yazan Haddad et al.
Journal of nanobiotechnology, 18(1), 95-95 (2020-07-15)
Currently, the diagnosis and treatment of neuroblastomas-the most frequent solid tumors in children-exploit the norepinephrine transporter (hNET) via radiolabeled norepinephrine analogs. We aim to develop a nanomedicine-based strategy towards precision therapy by targeting hNET cell-surface protein with hNET-derived homing peptides.
Jennifer E Gains et al.
European journal of nuclear medicine and molecular imaging, 45(3), 402-411 (2017-10-19)
Neuroblastoma may be treated with molecular radiotherapy, 131I meta-Iodobenzylguanidine and 177Lu Lutetium DOTATATE, directed at distinct molecular targets: Noradrenaline Transporter Molecule (NAT) and Somatostatin Receptor (SSTR2), respectively. This study used immunohistochemistry to evaluate target expression in archival neuroblastoma tissue, to
Isabel Pérez-Santos et al.
Cerebral cortex (New York, N.Y. : 1991), 31(9), 4115-4139 (2021-05-19)
Noradrenaline (NA) in the thalamus has important roles in physiological, pharmacological, and pathological neuromodulation. In this work, a complete characterization of NA axons and Alpha adrenoceptors distributions is provided. NA axons, revealed by immunohistochemistry against the synthesizing enzyme and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service