Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB5520-I

Sigma-Aldrich

Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, clone 7.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

7.1, monoclonal

mol wt

calculated mol wt 250.54 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Chondroitin sulfate proteoglycan 4 (UniProt: Q6UVK1; also known as Chondroitin sulfate proteoglycan NG2, Melanoma chondroitin sulfate proteoglycan, Melanoma-associated chondroitin sulfate proteoglycan) is encoded by the CSPG4 (also known as MCSP) gene (Gene ID: 1464) in human. Chondroitin sulfate proteoglycan NG2 is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-29), which is subsequently cleaved off to produce the mature form that contains a large extracellular domain (aa 30-2224), a transmembrane domain (aa 2225-2245), and a cytoplasmic domain (aa 2246-2322). NG2 plays a role in cell proliferation and migration and stimulates endothelial cells motility during microvascular morphogenesis. It is present in neural cells and in extraneural tissues, especially in the developing mesenchyme. Its level of expression is highest on immature, proliferating cells and decreases when these cells begin to differentiate. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells. It is a major chondroitin sulfate proteoglycan that is produced after spinal cord injury and is expressed by macrophages and oligodendrocyte progenitors. It is also reported to inhibit neurite outgrowth and growth cone collapse during axon regeneration.

Specificity

Clone 7.1 is a mouse monoclonal antibody that detects NG2 Chondroitin sulfate proteoglycan.

Immunogen

Human bone marrow stromal cells infected with SV-40.

Application

Quality Control Testing

Evaluated by Flow Cytometry in A375 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected NG2 Chondroitin Sulfate Proteoglycan in one million A375 cells.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated NG2 Chondroitin Sulfate Proteoglycan in Immunoprecipitation applications (Smith, F.O., et al. (1996). Blood;87(3):1123-33).

Flow Cytometry Analysis: A representative lot detected NG2 Chondroitin Sulfate Proteoglycan in Flow Cytometry applications (Morozov, A., et al. (2010). Clin Cancer Res.;16(23):5630-40; Behm, F.G., et al. (1996). Blood;87(3):1134-9; Smith, F.O., et al. (1996). Blood;87(3):1123-33).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-NG2 Chondroitin Sulfate Proteogycan, clone 7.1, Cat. No. MAB5520-I, is a mouse monoclonal antibody that detects NG2 Chondroitin sulfate proteoglycan and is tested for use in Flow Cytometry and Immunoprecipitation.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service